Skip to Content
Merck
All Photos(1)

Documents

C5914

Sigma-Aldrich

Cell Dissociation Solution Non-enzymatic 1x

Prepared in phosphate buffered saline without calcium and magnesium, sterile-filtered, BioReagent, suitable for cell culture

Synonym(s):

Dissociation Solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.75

Quality Level

sterility

sterile-filtered

product line

BioReagent

form

liquid

concentration

1 ×

technique(s)

cell culture | mammalian: suitable

pH

6.9-7.1

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Cell dissociation by non-enzymatic methods in human mesenchymal stromal cells aid in their detachment and preserves their functionality especially the expression of cell surface chemokine receptor.

Application

Cell Dissociation Solution Non-enzymatic 1x has been used for the dissociation of retinal astrocytes culture, U87 glioblastoma tumor cells and cardiac stem cells (CSCs).
Special non-enzymatic formulation for gently dislodging adherent cell types from plastic or glass surfaces.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ineke D C Jansen et al.
International journal of molecular sciences, 21(17) (2020-08-29)
Lysosome associated membrane proteins (LAMPs) are involved in several processes, among which is fusion of lysosomes with phagosomes. For the formation of multinucleated osteoclasts, the interaction between receptor activator of nuclear kappa β (RANK) and its ligand RANKL is essential.
Non-enzymatic dissociation of human mesenchymal stromal cells improves chemokine-dependent migration and maintains immunosuppressive function
Garg A, et al.
Cytotherapy, 16(4), 545-559 (2014)
Immunoimaging of CXCR4 expression in brain tumor xenografts using SPECT/CT
Nimmagadda S, et al.
Journal of Nuclear Medicine, 50(7), 1124-1130 (2009)
Isolation and characterization of murine retinal astrocytes
Scheef E, et al.
Molecular Vision, 11(4), 613-624 (2005)
Human cardiac stem cells with reduced notch signaling show enhanced therapeutic potential in a rat acute infarction model
Matsuda T, et al.
Circulation Journal, 78(1), 222-231 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service