Skip to Content
Merck
All Photos(1)

Documents

C3921

Sigma-Aldrich

Creatinase from Pseudomonas sp.

recombinant, expressed in E. coli, lyophilized powder, 10-15 units/mg protein

Synonym(s):

Creatine Amidinohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

form

lyophilized powder

specific activity

10-15 units/mg protein

mol wt

45 kDa

composition

Protein, ~70% biuret

storage temp.

2-8°C

Application

Creatine amidinohydrolase is a clinically important enzyme used in the determination of creatinine in blood and urine.

Biochem/physiol Actions

Creatinase is a homodimer with subunit molecular weights of approximately 45 kDa. It catalyzes the hydrolysis of creatine, in the presence of H2O, to form urea and sarcosine.

Unit Definition

One unit will hydrolyze 1.0 μmole of creatine to urea and sarcosine per min at pH 7.5 at 37 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Nakamura et al.
Bioscience, biotechnology, and biochemistry, 70(10), 2363-2370 (2006-10-13)
Aspergillus nidulans possessed 16 putative amylolytic genes consisting of 7 alpha-glucosidase (agdA-F), 7 alpha-amylase (amyA-F), and 2 glucoamylase (glaA and B) genes on the genome. Among them, the agdA, agdB, agdE, agdF, amyA, amyB, amyF, and glaB genes were induced
Pinar Esra Erden et al.
Artificial cells, blood substitutes, and immobilization biotechnology, 34(2), 223-239 (2006-03-16)
A new enzyme electrode for the determination of creatine was developed by immobilizing creatinase (CI) and sarcosine oxidase (SO). The enzymes were co-immobilized in a poly(vinylferrocenium) matrix onto the surface of a platinum working electrode. Crosslinking with glutaraldehyte (GA) and
Agnieszka Dzikowska et al.
Fungal genetics and biology : FG & B, 38(2), 175-186 (2003-03-07)
The arginine catabolism gene otaA encoding ornithine transaminase (OTAse) is specifically induced by arginine and is under the control of the broad-domain carbon and nitrogen repression systems. Arginine induction is mediated by a product of arcA gene coding for Zn(2)C(6)
N A Siddiqi et al.
Saudi journal of kidney diseases and transplantation : an official publication of the Saudi Center for Organ Transplantation, Saudi Arabia, 17(2), 203-207 (2006-08-15)
Polyoma virus nephropathy (BK virus) is being recognized as an important cause of graft failure. It is usually confused with acute rejection. No cases have been reported from the kingdom of Saudi Arabia. We report a case of a Saudi
M-L Lee
Pediatric cardiology, 26(6), 792-796 (2005-08-06)
I report on a 3-month-old infant with pulmonary atresia-intact ventricular septum and ventriculocoronary communication (VCC) who underwent percutaneous radiofrequency valvulotomy and valvuloplasty (RFVV). The patient's cardiac troponin-I, creatine kinase (CK), and myocardial fraction of (CK-MB) were elevated before RFVV and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service