Skip to Content
Merck
All Photos(1)

Key Documents

C2866

Sigma-Aldrich

Anti-Cannabinoid Receptor 1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

availability

not available in Japan

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 8-11 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

human ... CNR1(1268)
mouse ... Cnr1(12801)
rat ... Cnr1(25248)

Looking for similar products? Visit Product Comparison Guide

General description

Cannabinoid receptor type 1 (CNR1) is a G protein-coupled cannabinoid receptor found in central and peripheral nervous system. CNR1 belongs to rhodopsin-like G protein-coupled receptor superfamily.

Immunogen

synthetic peptide corresponding to the third cytoplasmic loop of human cannabinoid receptor 1, conjugated to KLH. The immunizing peptide has 100% homology with the rat and mouse gene.

Application

Anti-Cannabinoid Receptor 1 antibody produced in rabbit is suitable for immunocytochemistry and immunohistochemistry using formalin-fixed and paraffin-embedded sections at a concentration of 8-11μg/mL.

Biochem/physiol Actions

CNR1 has implications in several disease states, including drug addiction, anxiety, depression, obesity, and chronic pain. Different classes of CNR1 agonists are involved in signaling pathways through the activation of specific subtypes of G proteins. It may play a key role for the treatment of negative affective state produced by alcohol withdrawal and abstinence which is critical for the maintenance of alcohol addiction. CNR1 genes also play some roles in common polymorphism (rs1049353) with potential implications in weight loss.

Physical form

Solution in phosphate buffered saline, pH 7.7, containing 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue Shen et al.
BioMed research international, 2019, 5468954-5468954 (2019-02-26)
Adenomyosis is a common gynecologic benign disease that may have a life-long negative impact on women. Previous studies have indicated that the endocannabinoid system may participate in the progress of endometriosis. Our research aims to analyze the expression patterns of
Daniel Antonio de Luis et al.
Journal of investigative medicine : the official publication of the American Federation for Clinical Research, 62(2), 324-327 (2013-12-11)
A polymorphism (1359 G/A) of the cannabinoid receptor 1 (CNR1) gene was reported as a common polymorphism (rs1049353) with potential implications in weight loss. We decide to investigate the role of this polymorphism on metabolic changes and weight loss secondary
Joong-Youn Shim et al.
The Journal of biological chemistry, 288(45), 32449-32465 (2013-10-05)
The cannabinoid (CB1) receptor is a member of the rhodopsin-like G protein-coupled receptor superfamily. The human CB1 receptor, which is among the most expressed receptors in the brain, has been implicated in several disease states, including drug addiction, anxiety, depression
Jenny Ceccarini et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(8), 2822-2831 (2014-02-21)
Involvement of the type 1 cannabinoid receptor (CB1R) in the effects of alcohol on the brain is supported by animal experiments, but how in vivo CB1R levels are altered in alcoholic patients is still unclear. To assess the short-time effects
Özge Boyacıoğlu et al.
Cell death & disease, 12(1), 56-56 (2021-01-13)
Therapeutic agents used for non-small cell lung cancer (NSCLC) have limited curative efficacy and may trigger serious adverse effects. Cannabinoid ligands exert antiproliferative effect and induce apoptosis on numerous epithelial cancers. We confirmed that CB1 receptor (CB1R) is expressed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service