Skip to Content
Merck
All Photos(1)

Key Documents

C2780

Sigma-Aldrich

Chondroitinase AC from Flavobacterium heparinum

lyophilized powder, 0.5-1.5 units/mg protein (using chondroitin sulfate A as substrate)

Synonym(s):

Chondroitin AC lyase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

conjugate

(Glucosaminoglycan)

Quality Level

form

lyophilized powder

specific activity

0.5-1.5 units/mg protein (using chondroitin sulfate A as substrate)

composition

Protein, ~15% Lowry

solubility

0.02 M phosphate buffer: soluble (pH 7.0)

foreign activity

Glycosaminoglycan (GAG) degradation enzymes, may contain trace amount

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Chondroitinase AC from Sigma has been used for the large scale preparation of glycosaminoglycan (GAG) fractions during the study of structural and sequence motifs in dermatan sulfate.
Chondroitinase AC has been applied to the analysis of chondroitin sulfate in commercial samples such as dietary supplements.

Biochem/physiol Actions

Chondroitinase AC from Flavobacterium heparinum is an enzyme that cleaves sulfated and non-sulfated polysaccharide chains with (1-4) linkages between hexosamines and glucuronic acid residues, by an elimination mechanism. The resulting oligosaccharide products are mainly disaccharides with unsaturated uronic acids. Chondroitinase AC specifically degrades chondroitin sulfates A and C, but not chondroitin sulfate B (dermatan sulfate).

Other Notes

Contains potassium phosphate buffer salts and BSA as stabilizer.

Suitability

Depending on reaction conditions, digestion of chondroitin sulfate may result in hydrolysis of disaccharides

Unit Definition

One unit will cause a ΔA232 of 1.0 per minute due to the release of unsaturated disaccharide from chondroitin sulfate A at pH 7.3 at 37 °C. Reaction volume: 3.1 ml (light path 1 cm).

Preparation Note

Reconstitute the product in 20 mM phosphate buffer, pH 7.0. Subsequent dilutions can be made with a 0.01% aqueous bovine serum albumin solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Florian D Naal et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 87(1), 19-25 (2008-03-25)
Meniscal allograft processing procedures, in particular gamma irradiation, deteriorate the biomechanical and biological properties of the transplanted tissue. High hydrostatic pressure (HHP) treatment, widely used in food technology to inactivate microorganisms while preserving natural compounds, might serve as a gentle
S P Levine et al.
Blood, 75(4), 902-910 (1990-02-15)
Platelet factor 4 (PF4) is a hydrophobic, alpha-granule protein with potent antiheparin activity. It also binds to a chondroitin sulfate-containing proteoglycan (PG) isolated from platelets. In order to evaluate further the relationship between PF4 and the chondroitin sulfate-containing proteoglycan in
C B Knudson et al.
The Journal of cell biology, 100(5), 1753-1758 (1985-05-01)
Hyaluronate levels change dramatically during morphogenesis of various tissues and organs. Morphological detection of the exact temporal and spatial distribution patterns of hyaluronate may help to elucidate its role in morphogenesis. Since no specific direct method for visualizing hyaluronate with
Nicola Volpi et al.
Glycobiology, 19(4), 356-367 (2008-12-06)
Glycosaminoglycans from the body of marine clam Scapharca inaequivalvis were extracted at about 0.15- 0.18 mg/g of dry tissue, composed of dermatan sulfate (DS) (approx. 74%) and heparan sulfate (26%). After treatment with nitrous acid, DS was isolated for further
E B Hunziker
Osteoarthritis and cartilage, 9(1), 22-32 (2001-02-17)
We have previously shown (Hunziker and Rosenberg, J Bone Joint Surg 1996;78A:721-33) that synovial cells can be induced to migrate into partial-thickness articular cartilage defects, therein to proliferate and subsequently to deposit a scar-like tissue. We now wished to ascertain

Articles

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service