Skip to Content
Merck
All Photos(1)

Key Documents

C2571

Sigma-Aldrich

Anti-Human IgG (Fc specific)-Cy3 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

CY3 conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Application

Anti-Human IgG (Fc specific)-Cy3 antibody produced in goat was used to screen sera on glycopeptide microarrays at a dilution of 1:1000. It was used at a working dilution of 1:100 for immunohistochemistry analysis of mouse footpad sections incubated with human polyclonal anti-Leishmania serum.

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections. The coupling of Cy3 to Anti-Human IgG (Fc specific) antibody allows for the visualization of proteins by fluorescent microscopy.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Legal Information

Cy is distributed under license from Amersham Biosciences Limited.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jed Paul et al.
Journal of immunological methods, 432, 57-64 (2016-02-24)
Enzyme-linked immunosorbent assays (ELISAs) have traditionally been used to detect alloantibodies in patient plasma samples post hematopoietic cell transplantation (HCT); however, protein microarrays have the potential to be multiplexed, more sensitive, and higher throughput than ELISAs. Here, we describe the
Ola Blixt et al.
Breast cancer research : BCR, 13(2), R25-R25 (2011-03-10)
Detection of serum biomarkers for early diagnosis of breast cancer remains an important goal. Changes in the structure of O-linked glycans occur in all breast cancers resulting in the expression of glycoproteins that are antigenically distinct. Indeed, the serum assay
Jeffrey S Burgdorf et al.
Molecular psychiatry, 28(3), 1101-1111 (2022-12-10)
We developed an IGFBP2-mimetic peptide fragment, JB2, and showed that it promotes basal synaptic structural and functional plasticity in cultured neurons and mice. We demonstrate that JB2 directly binds to dendrites and synapses, and its biological activity involves NMDA receptor
Anna Chuprin et al.
Genes & development, 27(21), 2356-2366 (2013-11-05)
Cellular senescence limits proliferation of potentially detrimental cells, preventing tumorigenesis and restricting tissue damage. However, the function of senescence in nonpathological conditions is unknown. We found that the human placental syncytiotrophoblast exhibited the phenotype and expressed molecular markers of cellular
Aishun Jin et al.
Nature protocols, 6(5), 668-676 (2011-04-30)
Here we report a new method for isolating antigen-specific antibody-secreting cells (ASCs) using a microwell array chip, which offers a rapid, efficient and high-throughput (up to 234,000 individual cells) system for the detection and retrieval of cells that secrete antibodies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service