Skip to Content
Merck
All Photos(1)

Key Documents

C2389

Sigma-Aldrich

Cerulenin

from Cephalosporium caerulens, ≥98% (HPLC), powder, de novo phospholipid synthesis inhibitor

Synonym(s):

Helicocerin, (2R,3S,E,E)-2,3-Epoxy-4-oxo-7,10-dodecadienamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO3
CAS Number:
Molecular Weight:
223.27
Beilstein:
4140423
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Cerulenin, ≥98% (HPLC), from Cephalosporium caerulens

biological source

Cephalosporium caerulens

Quality Level

Assay

≥98% (HPLC)

form

powder

mp

93 °C

solubility

acetone: 19.60-20.40 mg/mL, clear to slightly hazy, colorless to yellow

antibiotic activity spectrum

fungi

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

C/C=C/C/C=C/CCC([C@@H]1[C@H](C(N)=O)O1)=O

InChI

1S/C13H18O3/c1-3-4-5-6-7-8-9-11(15)13-12(16-13)10(2)14/h3-4,6-7,12-13H,5,8-9H2,1-2H3/b4-3+,7-6+/t12-,13+/m0/s1

InChI key

PTNNGEBMCNMENY-JIVMHGEESA-N

Gene Information

human ... FASN(2194)

Looking for similar products? Visit Product Comparison Guide

General description

Cerulenin, an inhibitor of de novo phospholipid synthesis[1] triggers apoptosis in various wild type p53 and mutant p53 tumor cell lines.[2] This fungal antibiotic also blocks the condensing enzyme involved in fatty acid and polyketide biosynthesis.[3]

Application

Cerulenin has been used:

  • as a fatty acid synthase inhibitor to study its effects on aldosterone-induced trained immunity[4]
  • as a blocker of fatty acid synthase to study its effects on the replication of severe acute respiratory syndrome coronavirus 2 (SARS-CoV2)[5]
  • as a supplement in yeast extract–peptone–dextrose/glycerol (YPD/G) agar plates for the isolation of cerulenin-resistant yeast strains[6]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer W Lou et al.
Scientific reports, 9(1), 12987-12987 (2019-09-12)
During fatty acid biosynthesis, acyl carrier proteins (ACPs) from type I fungal fatty acid synthase (FAS) shuttle substrates and intermediates within a reaction chamber that hosts multiple spatially-fixed catalytic centers. A major challenge in understanding the mechanism of ACP-mediated substrate
Su Gao et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 301(1), R209-R217 (2011-04-22)
Hypothalamic fatty acid metabolism is involved in central nervous system controls of feeding and energy balance. Malonyl-CoA, an intermediate of fatty acid biosynthesis, is emerging as a significant player in these processes. Notably, hypothalamic malonyl-CoA has been implicated in leptin's
Biosynthesis of Enediyne Antibiotics
Iwasaki S
Chemistry & Biology (1999)
Darren C J Wong et al.
Frontiers in plant science, 9, 839-839 (2018-07-05)
Hundreds of orchid species secure pollination by sexually luring specific male insects as pollinators by chemical and morphological mimicry. Yet, the biochemical pathways involved in the synthesis of the insect sex pheromone-mimicking volatiles in these sexually deceptive plants remain poorly
Nepovirus
Sanfacon H, et al.
Encyclopedia of Virology (2008)

Articles

Cholesterol synthesis regulation by dietary levels, LDL receptors control lipid-rich LDL particle transport in cells.

Cholesterol synthesis regulation by dietary levels, LDL receptors control lipid-rich LDL particle transport in cells.

Cholesterol synthesis regulation by dietary levels, LDL receptors control lipid-rich LDL particle transport in cells.

Cholesterol synthesis regulation by dietary levels, LDL receptors control lipid-rich LDL particle transport in cells.

See All

Questions

  1. HOW LONG IS CERULENIN STABLE FOR WHEN DISSOLVED IN ACETONE?

    1 answer
    1. The solution stability of this product has not been determined. However, an identical product, 219557, notes that solutions prepared in organic solvents, such as acetone, may be stored protected from light at -20°C for up to 3 months. It is recommended that aqueous solutions not be stored for more than 1 day.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service