Skip to Content
Merck
All Photos(1)

Key Documents

C223

Sigma-Aldrich

Cirazoline hydrochloride

solid

Synonym(s):

2-[(2-Cyclopropylphenoxy)methyl]-4,5-dihydro-1H-imidazole hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N2O · HCl
CAS Number:
Molecular Weight:
252.74
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

form

solid

Quality Level

color

white

solubility

H2O: >35 mg/mL

SMILES string

Cl[H].C1CN=C(COc2ccccc2C3CC3)N1

InChI

1S/C13H16N2O.ClH/c1-2-4-12(11(3-1)10-5-6-10)16-9-13-14-7-8-15-13;/h1-4,10H,5-9H2,(H,14,15);1H

InChI key

XFRXWLWUUDJHPB-UHFFFAOYSA-N

Gene Information

Application

Cirazoline hydrochloride has been used to activate the α1-noradrenergic receptors and to study the inhibitory effect of neurokinin1 (NK1) antagonists on respiratory activity using the brainstem transverse slice.[1]

Biochem/physiol Actions

Cirazoline is a selective α1-adrenoceptor agonist and also a non-selective ligand for the imidazoline binding site.[2] Cirazoline dereases spatial working memory performance in aged monkeys.[3]

Features and Benefits

This compound is featured on the α1-Adrenoceptors and Imidazoline Binding Sites pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinhwan Kim et al.
Molecules (Basel, Switzerland), 24(4) (2019-02-24)
Although melanin production is a key self-defense mechanism against ultraviolet radiation (UVR)-induced skin damage, uneven or excessive deposition of melanin causes hyperpigmentary disorders. Currently available whitening agents are unsatisfactory because of issues with efficacy and safety. To develop more effective
Karen M Alsene et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 31(10), 2150-2161 (2006-01-13)
Prepulse inhibition (PPI) refers to the attenuation of startle when a weak prestimulus precedes the startling stimulus. PPI is deficient in several psychiatric illnesses involving poor sensorimotor gating. Previous studies indicate that alpha1 adrenergic receptors regulate PPI, yet the extent
Vito Bellantuono et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 148(2), 160-164 (2008-06-12)
Frog skin transports ions and water under hormonal control. In spite of the fundamental role played by adrenergic stimulation in maintaining the water balance of the organism, the receptor subtype(s) present in the skin have not been identified yet. We
A F Arnsten et al.
Pharmacology, biochemistry, and behavior, 58(1), 55-59 (1997-09-01)
The alpha-1 adrenergic agonist, cirazoline, was examined for effects on spatial working memory performance in aged rhesus monkeys. Cirazoline has additional high affinity for imidazoline receptors and has good brain penetrance when administered systemically. Spatial working memory was assessed using
Bronwyn A Evans et al.
Molecular pharmacology, 79(2), 298-307 (2010-10-28)
Although G protein-coupled receptors are often categorized in terms of their primary coupling to a given type of Gα protein subunit, it is now well established that many show promiscuous coupling and activate multiple signaling pathways. Furthermore, some agonists selectively

Articles

Review alpha 1 adrenoceptors as well as their agonists, antagonists, and tissue expression patterns. We suggest several modulators and alternatives for working with a-1 adrenoreceptors.

Review alpha 1 adrenoceptors as well as their agonists, antagonists, and tissue expression patterns. We suggest several modulators and alternatives for working with a-1 adrenoreceptors.

Review alpha 1 adrenoceptors as well as their agonists, antagonists, and tissue expression patterns. We suggest several modulators and alternatives for working with a-1 adrenoreceptors.

Review alpha 1 adrenoceptors as well as their agonists, antagonists, and tissue expression patterns. We suggest several modulators and alternatives for working with a-1 adrenoreceptors.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service