Skip to Content
Merck
All Photos(1)

Key Documents

C-22221

Sigma-Aldrich

Endothelial Cell Growth Medium MV 2

Basal Medium, 500 ml

Synonym(s):

Primary Cell Culture Media, Recommended with C-39226 or C-39221

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
Pricing and availability is not currently available.

form

aqueous solution

packaging

pkg of 500 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Endothelial Cell Basal Medium MV 2 (supplements sold separately)

To form a complete medium, it should be combined with the media supplements: C-39226 or C-39221

Application

Endothelial Cell Growth Medium MV2 is a low-serum (5% V/V) medium developed for the in vitro cultivation of endothelial cells from microvascular vessels, the coronary artery, and the aorta. The medium lacks the Endothelial Cell Growth Supplement (ECGS, bovine hypothalamic extract) but contains basic Fibroblast Growth Factor, Insulin-like Growth Factor and Vascular Endothelial Growth Factor (VEGF). Because of its effects on cell metabolism, VEGF may interfere with certain experimental setups. In these cases, we recommend the use of Endothelial Cell Growth Medium MV. The media are optimized for primary human cells but can also be used for bovine, porcine, murine, rat and dog endothelial cells from microvascular vessels as well as for various endothelial cell lines.Recommended for:Human Dermal Microvascular Endothelial Cells (HDMEC)Human Coronary Artery Endothelial Cells (HCAEC)Human Aortic Endothelial Cells (HAoEC)Human Pulmonary Microvascular Endothelial Cells (HPMEC)Human Dermal Lymphatic Endothelial Cells (HDLEC)Human Dermal Blood Endothelial Cells (HDBEC)Human Cardiac Microvascular Endothelial Cells (HCMEC)Human Uterine Microvascular Endothelial Cells (HUtMEC)Note: For best transfection results we recommend using PromoCell heparin-free Endothelial Cell Growth Medium MV2. Heparin, included all our other our Endothelial Cell Growth Media formulations, may reduce transfection efficiency.

Components

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials

Quality

All lots of PromoCell Endothelial Cell Media MV/MV2 are subjected to comprehensive quality control tests using primary human endothelial cells. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested endothelial cells. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Storage and Stability

Store the Basal Medium at 4 to 8°C in the dark immediately after arrival. Do not freeze the Basal Medium. If stored properly, the medium is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C

Subculture Routine

Click here for more information.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service