Skip to Content
Merck
All Photos(1)

Key Documents

C1493

Sigma-Aldrich

Cilnidipine

≥98% (HPLC), powder

Synonym(s):

1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid 2-methoxyethyl (2E)-3-phenyl-2-propenyl ester, FRC 8653

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H28N2O7
CAS Number:
Molecular Weight:
492.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

light yellow

solubility

H2O: ≤2 mg/mL
DMSO: ≥20 mg/mL

SMILES string

COCCOC(=O)C1=C(C)NC(C)=C(C1c2cccc(c2)[N+]([O-])=O)C(=O)OC\C=C\c3ccccc3

InChI

1S/C27H28N2O7/c1-18-23(26(30)35-14-8-11-20-9-5-4-6-10-20)25(21-12-7-13-22(17-21)29(32)33)24(19(2)28-18)27(31)36-16-15-34-3/h4-13,17,25,28H,14-16H2,1-3H3/b11-8+

InChI key

KJEBULYHNRNJTE-DHZHZOJOSA-N

Application

Cilnidipine has been used:
  • in cell viability assay of pheochromocytoma (nPC12) cells
  • in photoirradiation and photodegradation studies
  • to understand effect on albuminuria in diabetic nephropathy

Biochem/physiol Actions

A slow-acting Ca2+ channel blocker, antihypertensive; vasodilator.
Cilnidipine is a slow-acting Ca2+ channel blocker; antihypertensive; vasodilator; dual blocker of L-type voltage-gated Ca2+ channels in vascular smooth muscle and N-type Ca2+ channels in sympathetic nerve terminals that supply blood vessels. Cilnidipine may offer an advantage over nifedipine as the long term intake of the latter has been linked to increased risk of myocardial infarction and mortality in patients with coronary artery disease. Cilnidipine lowers blood pressure, but has less effect on sympathetic activity. Unlike nifedipine, cilnidipine does not inhibit PKC.

Features and Benefits

This compound is featured on the Calcium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Packaging

Protect from high temperatures.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sayaka Masada et al.
Scientific reports, 9(1), 11852-11852 (2019-08-16)
In July 2018, certain valsartan-containing drugs were voluntary recalled in Japan owing to contamination with N-nitrosodimethylamine (NDMA), a probable human carcinogen. In this study, an HPLC method was developed for the quantitative detection of NDMA simultaneously eluted with valsartan. Good
Cilnidipine mediates a neuroprotective effect by scavenging free radicals and activating the phosphatidylinositol 3-kinase pathway
Lee YJ, et al.
Journal of Neurochemistry, 111(1), 90-100 (2009)
Photoinduced Aromatization of Asymmetrically Substituted 1, 4-Dihydropyridine Derivative Drug Cilnidipine
Ahmad W
Journal of Photochemistry, 2014(1), 90-100 (2014)
Kazuomi Kario et al.
Journal of clinical hypertension (Greenwich, Conn.), 15(2), 133-142 (2013-01-24)
The Ambulatory Blood Pressure Control and Home Blood Pressure (Morning and Evening) Lowering By N-Channel Blocker Cilnidipine (ACHIEVE-ONE) trial is a large-scale clinical study on blood pressure (BP) and pulse rate (PR) in the real world with use of cilnidipine
Liandong Hu et al.
Carbohydrate polymers, 90(4), 1719-1724 (2012-09-05)
The objective of this study was to improve the water-solubility and photostability of cilnidipine by complexing it with hydroxypropyl-β-cyclodextrin (HP-β-CD or HP-beta-CD). The interactions of cilnidipine and HP-β-CD were characterized by ultra violet-visible (UV/VIS) spectroscopy, differential scanning calorimetry (DSC), powder

Articles

We offers many products related to calcium channels for your research needs.

We offers many products related to calcium channels for your research needs.

We offers many products related to calcium channels for your research needs.

We offers many products related to calcium channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service