Skip to Content
Merck
All Photos(1)

Key Documents

C-12302

Sigma-Aldrich

Normal Human Dermal Fibroblasts (NHDF)

Adult, 500,000 cryopreserved cells

Synonym(s):

NHDF cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81
Pricing and availability is not currently available.

biological source

human dermis

packaging

pkg of 500,000 cells

morphology

(fibroblast)

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Cell Line Origin

Skin

Application

Primary Normal Human Dermal Fibroblasts (NHDF) are isolated form the dermis of adult skin (different locations) and are provided in a cryopreserved format. They can be used for wound healing studies and dermatological research to investigate diseases like scleroderma, fibrosarcoma, fibrosis, xeroderma pigmentosum, and histiocytoma. Moreover, fibroblasts are important for cancer research, tissue regeneration, and tissue engineering studies.Normal Human Epidermal Keratinocytes (NHEK) and Normal Human Epidermal Melanocytes (NHEM) cultured in M2 medium from the same donor are available on request.

Quality

Rigid quality control tests are performed for each lot of Human Fibroblasts. They are tested for cell morphology, adherence rate and cell viability. Flow cytometric analyses of the fibroblast specific antigen CD90 are performed for each lot. Growth performance is tested through multiple passages up to 10 or 15 population doublings (PD) under culture conditions without antibiotics and antimycotics. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Subculture Routine

Click here for more information.

Other Notes

Recommended Plating Density: 3500 - 7000 cells per cm2Passage After Thawing: P2Tested Markers: CD90 positiveGuaranteed population doublings: >15

Recommended products

Recommended Primary Cell Culture Media:Link
Recommended Primary Cell Culture Media:Link

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Yeast plasmid selection: Utilizes auxotrophic yeast strains for plasmid selection, enabling growth on synthetic media for better yield.

Yeast plasmid selection: Utilizes auxotrophic yeast strains for plasmid selection, enabling growth on synthetic media for better yield.

Yeast plasmid selection: Utilizes auxotrophic yeast strains for plasmid selection, enabling growth on synthetic media for better yield.

Yeast plasmid selection: Utilizes auxotrophic yeast strains for plasmid selection, enabling growth on synthetic media for better yield.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service