Skip to Content
Merck
All Photos(2)

Documents

C0791

Sigma-Aldrich

Monoclonal Anti-Cytokeratin Peptide 13 antibody produced in mouse

clone KS-1A3, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

KS-1A3, monoclonal

contains

15 mM sodium azide

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect immunofluorescence: 1:100 using formalin-fixed, paraffin-embedded human tissue sections.1
microarray: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT13(3860)

Specificity

The antibody reacts with squamous, non-keratinized epithelium, transitional epithelium, pseudostratified epithelium, and myoepithelium.

Immunogen

cultured human epidermoid carcinoma cell line A-431.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Monoclonal Anti-Cytokeratin Peptide 13 antibody produced in mouse is suitable for the following applications:
  • Immunohistochemistry (formalin-fixed, paraffin-embedded sections)
  • Indirect immunofluorescence at a working dilution of 1:100 using formalin-fixed, paraffin-embedded human tissue sections.
  • Microarray

Biochem/physiol Actions

Cytokeratins are proteins of keratin-containing intermediate filaments that provide mechanical support and other additional functions in epithelial cells. It is found in the intracytoplasmic cytoskeleton of epithelial tissue. Epithelial tissue expresses cytokeratin subunits in a specific and stable pattern. Cytokeratins along with vimentin are involved in cell proliferation, migration and differentiation of preodontoblasts and preameloblasts. The intermediate-sized filaments are abundant in human endothelial cells and are mostly of vimentin type.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huan He et al.
The Journal of biological chemistry, 290(21), 13567-13577 (2015-04-09)
Squamous cell differentiation requires the coordinated activation and repression of genes specific to the differentiation process; disruption of this program accompanies malignant transformation of epithelium. The exploration of genes that control epidermal proliferation and terminal differentiation is vital to better
Helen M Rigden et al.
PloS one, 11(5), e0156009-e0156009 (2016-05-27)
To quantify the extent of squamous metaplasia in bronchial biopsies and relate it to the presence of chronic obstructive pulmonary disease (COPD), a smoking-related pathology. Bronchial biopsies (n = 15 in each group) from smokers with COPD GOLD stage1 and
Debarchana Saha et al.
PloS one, 12(2), e0171084-e0171084 (2017-02-09)
Hemoglobin (Hb) is a major protein involved in transport of oxygen (O2). It consists of Hb-α and Hb-β subunits, which are normally expressed by cells of erythroid lineage. However, till recently, it was not known whether non-erythroid cells like vaginal
Aurélie Avril-Delplanque et al.
Stem cells (Dayton, Ohio), 23(7), 992-1001 (2005-07-27)
Airway epithelium stem cells have not yet been prospectively identified, but it is generally assumed that both secretory and basal cells have the capacity to divide and differentiate. Previously, we developed a test for progenitor cells of the human airway
S Spuler et al.
The American journal of pathology, 148(5), 1359-1365 (1996-05-01)
To study the possible role of thymomas and of extrathymomal thymic tissue in the development and maintenance of myasthenia gravis, we transplanted fragments of either tissue into SCID mice and monitored the production of anti-acetylcholine receptor antibodies in the recipients.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service