Key Documents
BMSIPS2050PT3
Sigma-Aldrich
Benchmark Overhead Stirrer Propeller
flat paddle with holes, PTFE-coated
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
material
PTFE-coated
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific IPS2050-P-T3
diam. × L
68 mm × 350 mm
General description
Stiring rods and propellers to fit Benchmark range of OS-20L and OS-40L overhead stirrers.
Useful for gentle, mid to low speed mixing, this PTFE coated flat propeller has 3 holes.
Application
stirring, mixing