BAB617003400
H-B® Durac® Timer
Timing capacity : 99 Minute:59 Second, digital
Synonym(s):
laboratory timer, pocket timer, timer alarm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.79
Recommended Products
feature
digital
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art B61700-3400
display W × H
48.3 mm × 15.2 mm
Looking for similar products? Visit Product Comparison Guide
General description
Single channel timer has three key operation for ease of use.
Features and Benefits
Timing capacity: 99 minutes: 59 seconds
Count direction: up/down
Last time set memory
Audible alarm; displays elapsed time after countdown
Mounting options: lay flat on bench, stand on edge for vertical view, magnetic back for metal surface attachment
Requires one AAA battery (included)
Count direction: up/down
Last time set memory
Audible alarm; displays elapsed time after countdown
Mounting options: lay flat on bench, stand on edge for vertical view, magnetic back for metal surface attachment
Requires one AAA battery (included)
Legal Information
Durac is a registered trademark of SP Scienceware, a division of SP
H-B is a registered trademark of SP Scienceware, a division of SP
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service