Skip to Content
Merck
All Photos(1)

Key Documents

B8438

Sigma-Aldrich

Alcian Blue solution

1% in 3% acetic acid, pH 2.5

Synonym(s):

Alcian Blue 8XG solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47
Pricing and availability is not currently available.

form

liquid

Quality Level

concentration

1% in 3% acetic acid, pH 2.5

technique(s)

microbe id | staining: suitable

color

dark blue to very dark blue

pH range

2.4-2.6

Amax

0.45-0.60 (1:400 in H2O)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

Application

Alcian Blue solution has been used in the histological staining of acidic glycosaminoglycans (GAGs; mucopolysaccharides) that are carboxylated or sulfated.[1][2] Sulfated glycosaminoglycans appear to be the preferred substrate for alcian blue at pH 2.5.[3] Alcian blue has been used to stain hyaluronan (HA), a nonsulfated GAG, but the intensity of the staining of HA is affected by the method of tissue fixation used.[3] Alcian blue staining of GAGs in tissue may be modified through the addition of an electrolyte such as magnesium chloride.[4]

Biochem/physiol Actions

Alcian Blue is a cationic dye. It forms insoluble complexes with acidic glycosaminoglycans, thereby aiding in the quantitative determination of glycosaminoglycans.[5] It is also used for the staining of bone and cartilage.[6]

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SPECIFIC STAINING OF SULPHATE GROUPS WITH ALCIAN BLUE AT LOW PH.
R LEV et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 12, 309-309 (1964-04-01)
A procedure for differential staining of cartilage and bone in whole formalin-fixed vertebrates.
Wassersug R J.
Stain Technology, 51(2), 131-134 (1976)
Hao Wu et al.
Experimental cell research, 361(2), 324-332 (2017-11-04)
Mesenchymal stem-cell based therapies have been proposed as novel treatments for intervertebral disc (IVD) degeneration. The development of these treatment strategies, however, has been hindered by the incomplete understanding of the origin, biological properties of nucleus pulposus (NP) derived stem/progenitor
G F Pierce et al.
The American journal of pathology, 138(3), 629-646 (1991-03-01)
Recombinant platelet-derived growth factor (PDGF) and transforming growth factor beta 1 (TGF-beta 1) influence the rate of extracellular matrix formed in treated incisional wounds. Because incisional healing processes are difficult to quantify, a full-thickness excisional wound model in the rabbit
Differential staining of acid glycosaminoglycans (mucopolysaccharides) by alcian blue in salt solutions.
J E Scott et al.
Histochemie. Histochemistry. Histochimie, 5(3), 221-233 (1965-10-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service