Skip to Content
Merck
All Photos(1)

Key Documents

B8436

Sigma-Aldrich

Anti-Brachyury antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-T, Anti-T Brachyury, mouse homolog, Anti-T brachyury-like, Anti-TFT, Anti-Transcription factor T

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

immunohistochemistry: 1:100-1:250
western blot: 1:500-1:1,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... T(6862)
mouse ... T(20997)
rat ... T(292301)

General description

Brachyury (T) is an oncogene mapped to human chromosome 6q27. This gene codes for Brachyury, which is expressed throughout the primary mesoderm during the early stages of embryogenesis. Brachyury protein structure comprises highly conserved DNA binding domain, known as T-box domain.

Immunogen

synthetic peptide corresponding to an internal sequence of human brachyury (GeneID 6862). This sequence is 80% identical in mouse and 86% identical in rat.

Biochem/physiol Actions

Brachyury protein is a transcription factor that aids in the embryonic development of notochord tissue. It also plays an essential role in posterior mesoderm formation and differentiation. Brachyury is highly expressed in a variety of cancers including, chordoma and hemangioblastoma. It serves as an immunohistochemical marker for differential diagnosis of chordoma and hemangioblastoma (HBL).

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott W Connors et al.
F1000Research, 9 (2020-07-30)
Chordomas are rare and difficult-to-treat tumors arising from the embryonic notochord. While surgery is the mainstay of treatment, and despite new techniques aimed at maximizing total tumoral resection, recurrence remains high and the probability of disease-free survival low. New breakthroughs
Ashley E E Bruce et al.
Mechanisms of development, 163, 103625-103625 (2020-06-12)
The Brachyury gene encodes a transcription factor that is conserved across all animals. In non-chordate metazoans, brachyury is primarily expressed in ectoderm regions that are added to the endodermal gut during development, and often form a ring around the site
Fabio Papes et al.
Nature communications, 13(1), 2387-2387 (2022-05-03)
Transcription Factor 4 (TCF4) has been associated with autism, schizophrenia, and other neuropsychiatric disorders. However, how pathological TCF4 mutations affect the human neural tissue is poorly understood. Here, we derive neural progenitor cells, neurons, and brain organoids from skin fibroblasts
Jianjian Zhu et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(14), 3820-3825 (2016-03-24)
The transcription factor Brachyury (T) gene is expressed throughout primary mesoderm (primitive streak and notochord) during early embryonic development and has been strongly implicated in the genesis of chordoma, a sarcoma of notochord cell origin. Additionally, T expression has been
Valeria Barresi et al.
Disease markers, 2014, 514753-514753 (2014-03-05)
Brachyury is a transcription factor which is required for posterior mesoderm formation and differentiation as well as for notochord development during embryogenesis. Due to its expression in the neoplastic cells of chordoma, a malignant tumour deriving from notochordal remnants, but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service