Skip to Content
Merck
All Photos(3)

Key Documents

B8378

Sigma-Aldrich

2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole

≥99%

Synonym(s):

2-(4-Biphenylyl)-5-(4-tert-butylphenyl)-1,3,4-oxadiazole, Butyl-PBD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H22N2O
CAS Number:
Molecular Weight:
354.44
Beilstein:
1086414
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥99%

form

powder

fluorescence

λex 305 nm; λem 364 nm in ethanol

OLED Device Performance

ITO/DTFD/PBD/Alq3/LiF/Al

  • Color: blue
  • Max. Luminance: 400 Cd/m2
  • Max. EQE: 1.6 %
  • Turn-On Voltage: 4 V

ITO/PEDOT:PSS/TPD/PVK:Ir(ppy)3 (6%)/PBD/Al
  • Color: green
  • Max. Luminance: 10000 Cd/m2

storage temp.

2-8°C

SMILES string

CC(C)(C)c1ccc(cc1)-c2nnc(o2)-c3ccc(cc3)-c4ccccc4

InChI

1S/C24H22N2O/c1-24(2,3)21-15-13-20(14-16-21)23-26-25-22(27-23)19-11-9-18(10-12-19)17-7-5-4-6-8-17/h4-16H,1-3H3

InChI key

XZCJVWCMJYNSQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole (PBD) can be used as a hole-blocking material in the fabrication of multilayer polymer light-emitting diodes to enhance the efficiency of the device. It is also used as an additive to fabricate the polyurethane (PU) based bistable nonvolatile memory devices with resistive switching characteristics. By incorporating PBD with PU, device performance showed significant improvement, with increase in ON/OFF ratio by two orders of magnitude and a long retention period of over five hours. PBD is also an organic fluorescent molecule used to prepare plastic scintillators.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective Metal Transfer and its Application to Patterned Multicolor Organic Light-Emitting Diodes
Lee, D.; et al.
Advanced Materials, 16, 1851 - 1854 (2011)
Blue-light-emitting organic electroluminescence via exciplex emission based on a fluorene derivative
Li, F.; et al.
Journal of Physics D: Applied Physics, 37, 1613-1613 (2004)
Multilayer polymer light-emitting diodes by blade coating method
Tseng S-R, et al.
Applied Physics Letters, 93(15), 382-382 (2008)
Development of polymerizable 2-(1-naphthyl)-5-phenyloxazole scintillators for ionizing radiation detection
Seliman AF, et al.
Journal of Material Chemistry C, 3(27), 7053-7061 (2015)
Bistable electrical switching characteristics and memory effect by mixing of oxadiazole in polyurethane layer
S Yanmei, et al.
The Journal of Physical Chemistry C, 119(33), 19520-19525 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service