Skip to Content
Merck
All Photos(1)

Key Documents

B7431

Sigma-Aldrich

Bryostatin 1

≥99%, solid

Synonym(s):

NSC 339555

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H68O17
CAS Number:
Molecular Weight:
905.03
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥99%

form

solid

color

white

solubility

DMSO: soluble
ethanol: soluble

storage temp.

−20°C

SMILES string

[H]\C(C(=O)OC)=C1/C[C@@]2([H])C[C@]3(O)O[C@]([H])(C[C@@H](O)CC(=O)O[C@H](C[C@]4([H])C\C(=C(\[H])C(=O)OC)[C@H](OC(=O)\C=C\C=C\CCC)[C@@](O)(O4)C(C)(C)\C=C\[C@@]([H])(C1)O2)[C@@H](C)O)C[C@H](OC(C)=O)C3(C)C

InChI

1S/C47H68O17/c1-10-11-12-13-14-15-39(51)62-43-31(22-41(53)58-9)21-34-25-37(28(2)48)61-42(54)24-32(50)23-35-26-38(59-29(3)49)45(6,7)46(55,63-35)27-36-19-30(20-40(52)57-8)18-33(60-36)16-17-44(4,5)47(43,56)64-34/h12-17,20,22,28,32-38,43,48,50,55-56H,10-11,18-19,21,23-27H2,1-9H3/b13-12+,15-14+,17-16+,30-20+,31-22+/t28-,32-,33+,34+,35-,36+,37-,38+,43+,46+,47-/m1/s1

InChI key

MJQUEDHRCUIRLF-TVIXENOKSA-N

Application

Bryostatin 1 has been used to study its effects on spontaneous Crohn′s disease (CD) like colitis in mice.[1] It has also been used to study its effects as an anthelmintic drug on adult Syphacia muris infection in rats.[2]

Biochem/physiol Actions

Bryostatin 1 (Bry1) is a macrocyclic lactone.[3] This synaptogenic compound[4] is obtained from the marine bryozoan Bugula neritina.[1] Bry1 is capable of reversing synaptic loss. It can enable synaptic maturation in animal models with several neurological disorders.[3] It possesses antidepressant activity, when administered intracebroventricularly.[4] Bry1 is known to participate in protecting cell tight junctions (TJs), anti-inflammatory functions and immune regulation.[1]
Macrolactone isolated from the marine bryozoan Bugula neritina that initially activates and subsequently down-regulates protein kinase C (PKC). More potent than phorbol myristate acetate for translocating PKCδ and ε.

Other Notes

Binds to glass and plastic in aqueous solutions

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Douglas Smith et al.
Leukemia research, 35(1), 87-94 (2010-07-06)
Pharmacologic differentiating agents have had relatively limited clinical success outside of the use of ATRA in acute promyelocytic leukemia and DNA methyltransferase inhibitors in myelodysplastic syndromes. The differentiating effects of such agents can be enhanced in combination with lineage-specific growth
Maria Eugenia Ariza et al.
The Journal of biological chemistry, 286(1), 24-34 (2010-11-03)
Bryostatin-1 (Bryo-1), a natural macrocyclic lactone, is clinically used as an anti-cancer agent. In this study, we demonstrate for the first time that Bryo-1 acts as a Toll-like receptor 4 (TLR4) ligand. Interestingly, activation of bone marrow-derived dendritic cells (in
Jarin Hongpaisan et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(2), 630-643 (2011-01-14)
Among the pathologic hallmarks of Alzheimer's disease (AD) neurodegeneration, only synaptic loss in the brains of AD patients closely correlates with the degree of dementia in vivo. Here, we describe a molecular basis for this AD loss of synapses: pathological
Maciej Kmieciak et al.
Journal of visualized experiments : JoVE, (47)(47), doi:10-doi:10 (2011-02-10)
It was reported that breast cancer patients have pre-existing immune responses against their tumors(1,2). However, such immune responses fail to provide complete protection against the development or recurrence of breast cancer. To overcome this problem by increasing the frequency of
Moisés Pérez et al.
Current HIV research, 8(6), 418-429 (2010-07-20)
The persistence of latent HIV-infected cellular reservoirs represents the major hurdle to virus eradication on patients treated with HAART. It has been suggested that successful depletion of such latent reservoirs will require a combination of therapeutic agents that can specifically

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service