Skip to Content
Merck

B6048

Restriction Endonuclease Buffer SP

2 ×

Synonym(s):

Restriction Endonuclease Buffer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Level

description

2× buffer contains 200 mM Tris-HCl, pH 7.6, 20 mM MgCl2, 300 mM NaCl, 2 mM dithiothreitol at 37 °C.

form

buffered aqueous solution

concentration

2 ×

storage temp.

−20°C

SMILES string

CC(C)CC(CNC(Cc1ccccc1)C(N)=O)NC(=O)C(Cc2cnc[nH]2)NC(=O)CNC(=O)C(NC(=O)C(C)NC(=O)C(Cc3c[nH]c4ccccc34)NC(=O)C(CCC(N)=O)NC(=O)C(N)Cc5c[nH]c6ccccc56)C(C)C

InChI

1S/C58H77N15O9/c1-32(2)21-39(29-65-46(52(61)76)22-35-13-7-6-8-14-35)69-57(81)48(25-38-28-62-31-67-38)70-50(75)30-66-58(82)51(33(3)4)73-53(77)34(5)68-56(80)47(24-37-27-64-44-18-12-10-16-41(37)44)72-55(79)45(19-20-49(60)74)71-54(78)42(59)23-36-26-63-43-17-11-9-15-40(36)43/h6-18,26-28,31-34,39,42,45-48,51,63-65H,19-25,29-30,59H2,1-5H3,(H2,60,74)(H2,61,76)(H,62,67)(H,66,82)(H,68,80)(H,69,81)(H,70,75)(H,71,78)(H,72,79)(H,73,77)

InChI key

YNZJHLFHFNCZIE-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
B6048-1VL04061833294703