Skip to Content
Merck
All Photos(1)

Key Documents

B5399

Sigma-Aldrich

(±)-Baclofen

≥98% (HPLC), solid

Synonym(s):

(±)-β-(Aminomethyl)-4-chlorobenzenepropanoic acid, Lioresal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12ClNO2
CAS Number:
Molecular Weight:
213.66
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98% (HPLC)

form

solid

color

white to very faintly yellow

solubility

1 M HCl: 50 mg/mL

storage temp.

2-8°C

SMILES string

ClC1=CC=C(C(CN)CC(O)=O)C=C1

InChI

1S/C10H12ClNO2/c11-9-3-1-7(2-4-9)8(6-12)5-10(13)14/h1-4,8H,5-6,12H2,(H,13,14)

InChI key

KPYSYYIEGFHWSV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

(±)-Baclofen has been used as γ-aminobutyric acid receptor B GABAB receptor agonist:
  • as well as control GABAergic drug to test its protective effects on 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP)-induced dopaminergic toxicity
  • to test its effects on in vivo motor function tests and performance of mutant mice
  • to test its excitability effect on dopaminergic (DA) neurons of the ventral tegmental area (VTA)
  • to test its effect in reducing dendritic excitability in Purkinje neurons

Biochem/physiol Actions

Baclofen, a γ-aminobutyric acid (GABA) analog, possesses myorelaxant properties Being a γ-aminobutyric acid receptor B (GABAB) agonist, baclofen may be involved in the potentiation of dendritic potassium Kchannels. It may be useful in blocking transient lower esophageal sphincter relaxation (TLESR) in gastroesophageal reflux disease (GERD). Baclofen may also have scope for treating addictive especially, in alcohol use disorder (AUD).
GABAB receptor agonist; skeletal muscle relaxant; antispastic agent.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Abhiram Pushparaj et al.
Behavioural brain research, 290, 77-83 (2015-05-03)
Our prior work demonstrated the involvement of the caudal granular subregion of the insular cortex in a rat model of nicotine self-administration. Recent studies in various animal models of addiction for nicotine and other drugs have identified a role for
The effects of baclofen for the treatment of gastroesophageal reflux disease: a meta-analysis of randomized controlled trials.
Li, et al.
Gastroenterology Research and Practice, 2014, 307805-307805 (2020)
The Use of Baclofen as a Treatment for Alcohol Use Disorder: A Clinical Practice Perspective.
de Beaurepaire, et al.
Frontiers in Psychiatry, 9, 708-708 (2021)
Mohamed-Ali Gorsane et al.
Substance abuse, 33(4), 336-349 (2012-09-20)
Baclofen, a γ-aminobutyric acid (GABA)-B receptor agonist, represents a promising drug in alcohol addiction management. Animal models have shown its action at various stages of the process of alcohol addiction. Moreover, initial open and randomized controlled trials have shown the
Andrés P Varani et al.
Psychopharmacology, 231(15), 3031-3040 (2014-02-21)
Nicotine is a major active ingredient in tobacco and plays a major role in tobacco addiction. In rodents, repeated nicotine administration produces behavioral responses related to its addictive properties, such as reinforcing effects and physical dependence. The aim of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service