Skip to Content
Merck
All Photos(1)

Documents

B4383

Sigma-Aldrich

Bergamot oil

Synonym(s):

Bergamot peel oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

refractive index

n20/D 1.4665 (lit.)
n20/D 1.4675 (lit.)

bp

159 °C (lit.)

density

0.877 g/mL at 25 °C (lit.)
0.88 g/mL at 25 °C (lit.)

lipid type

oils

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deborah H Ndao et al.
Psycho-oncology, 21(3), 247-254 (2012-03-03)
Though often lifesaving, stem cell transplantation (SCT) is a period of great distress for both child and parent. We conducted a double-blind, placebo-controlled randomized study evaluating the effect of the respiratory administration of bergamot essential oil on the anxiety, nausea
Tsukasa Sakurada et al.
Pharmacology, biochemistry, and behavior, 97(3), 436-443 (2010-10-12)
This study investigated the effect of bergamot essential oil (BEO) containing linalool and linalyl acetate as major volatile components in the capsaicin test. The intraplantar injection of capsaicin (1.6 μg) produced a short-lived licking/biting response toward the injected paw. The
Ji-Yeong Seo
Journal of Korean Academy of Nursing, 39(3), 357-365 (2009-07-03)
This study was done to examine the effects of aromatherapy on stress and stress responses in adolescents. A two-group cross-over design was used for this study. The experimental treatment was aroma essential oil inhalation and the placebo treatment was carrier
K Fisher et al.
Letters in applied microbiology, 44(5), 495-499 (2007-04-25)
To test the effect of oils and vapours of lemon, sweet orange and bergamot and their components against three Arcobacter butzleri strains. The disc diffusion method was used to screen the oils and vapours against three strains of A. butzleri.
M T Corasaniti et al.
British journal of pharmacology, 151(4), 518-529 (2007-04-03)
The effects of bergamot essential oil (BEO; Citrus bergamia, Risso) on excitotoxic neuronal damage was investigated in vitro. The study was performed in human SH-SY5Y neuroblastoma cells exposed to N-methyl-D-aspartate (NMDA). Cell viability was measured by dye exclusion. Reactive oxygen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service