Skip to Content
Merck
All Photos(1)

Key Documents

B2417

Sigma-Aldrich

Benzamil hydrochloride hydrate

≥98% (HPLC)

Synonym(s):

N-(Benzylamidino)-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14ClN7O · HCl · xH2O
CAS Number:
Molecular Weight:
356.21 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

yellow

solubility

methanol: 10 mg/mL
DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

O.Cl.Nc1nc(N)c(nc1Cl)C(=O)NC(=N)NCc2ccccc2

InChI

1S/C13H14ClN7O.ClH.H2O/c14-9-11(16)20-10(15)8(19-9)12(22)21-13(17)18-6-7-4-2-1-3-5-7;;/h1-5H,6H2,(H4,15,16,20)(H3,17,18,21,22);1H;1H2

InChI key

YAJQRPFIYXRNKS-UHFFFAOYSA-N

Biochem/physiol Actions

Selective and potent blocker of Na+/H+ and Na+/Ca2+ channels

Features and Benefits

This compound is featured on the Acid-Sensing (Proton-gated) Ion Channels (ASICs) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Other Notes

Nω-Benzyl derivative of amiloride.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G N Pierce et al.
The Journal of pharmacology and experimental therapeutics, 265(3), 1280-1291 (1993-06-01)
Amiloride and its derivatives (benzamil, dichlorobenzamil, 5-(N,N-dimethyl)-amiloride, 5-(N-ethyl-N-isopropyl)-amiloride, (N,N-hexamethylene)- amiloride and 5-(N-methyl-N-isobutyl)-amiloride) are commonly used as selective blockers of Na+/Ca++ exchange or Na+/H+ exchange. Very little information is currently available regarding their effects on cardiac performance. It was observed that
J W Lane et al.
British journal of pharmacology, 106(2), 283-286 (1992-06-11)
1. Patch clamp recording techniques have been used to compare the block caused by amiloride and some of its structural analogues of the mechanosensitive (MS) cation selective channel in frog (Xenopus laevis) oocytes. 2. Like amiloride, the amiloride analogues dimethylamiloride
A Catalina Vélez-Ortega et al.
eLife, 6 (2017-03-30)
Mechanotransducer channels at the tips of sensory stereocilia of inner ear hair cells are gated by the tension of 'tip links' interconnecting stereocilia. To ensure maximal sensitivity, tip links are tensioned at rest, resulting in a continuous influx of Ca2+
Sigrid Noreng et al.
eLife, 9 (2020-07-31)
The molecular bases of heteromeric assembly and link between Na+ self-inhibition and protease-sensitivity in epithelial sodium channels (ENaCs) are not fully understood. Previously, we demonstrated that ENaC subunits - α, β, and γ - assemble in a counterclockwise configuration when
Angela Ballesteros et al.
eLife, 7 (2018-08-01)
The hair cell mechanotransduction (MET) channel complex is essential for hearing, yet it's molecular identity and structure remain elusive. The transmembrane channel-like 1 (TMC1) protein localizes to the site of the MET channel, interacts with the tip-link responsible for mechanical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service