Skip to Content
Merck
All Photos(4)

Key Documents

B2156

Sigma-Aldrich

Benzethonium hydroxide solution

~1.0 M in methanol (by HCl titration)

Synonym(s):

Benzethonium, Benzethonium Hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H43NO3
CAS Number:
Molecular Weight:
429.64
Beilstein:
3915309
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

cationic

Quality Level

form

liquid

mol wt

429.64 g/mol

concentration

~1.0 M in methanol (by HCl titration)

technique(s)

microbiological culture: suitable

storage temp.

2-8°C

SMILES string

[OH-].CC(C)(C)CC(C)(C)c1ccc(OCCOCC[N+](C)(C)Cc2ccccc2)cc1

InChI

1S/C27H42NO2.H2O/c1-26(2,3)22-27(4,5)24-13-15-25(16-14-24)30-20-19-29-18-17-28(6,7)21-23-11-9-8-10-12-23;/h8-16H,17-22H2,1-7H3;1H2/q+1;/p-1

InChI key

USLZJBWCHRRIAQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Benzethonium hydroxide solution has been used for the trapping of 14CO2. Benzethonium hydroxide is suitable to evaluate the kinetics of 14carbon dioxide (14CO2) excretion by oral commensal flora to theoretically propose optimum breath collection timings for 14C urea breath test.

Biochem/physiol Actions

Benzethonium is a cationic of quaternary ammonium salt consisting of diisobutylphenoxyethoxyethyl dimethyl-benzyl ammonium compound. It possess antimicrobial and antiseptic functionalities. Benzethonium hydroxide conversion to carbamate in the presence of exhaled carbon dioxide results in colour change. Hence, it is used in carbon urea breath test.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmaceutical interference with the [14C] carbon urea breath test for the detection of Helicobacter pylori infection
Abrams DN, et al.
J. Pharm. Pharm. Sci., 3(2), 228-233 (2000)
Influence of the anions on the N-cationic benzethonium salts in the solid state and solution: Chloride, bromide, hydroxide and citrate hydrates
Paradies HH and Reichelt H
AIP Advances, 6(6), 065322-065322 (2016)
Wnt-induced activation of glucose metabolism mediates the in vivo neuroprotective roles of Wnt signaling in Alzheimer disease
Cisternas P, et al.
Journal of Neurochemistry, 149, 54?72-54?72 (2018)
Kinetics of 14carbon dioxide excretion from 14C-urea by oral commensal flora
Pathak CM, et al.
Journal of gastroenterology and hepatology, 23(10), 1603-1607 (2008)
Measurements of glucose conversion to its metabolites
Adipose Tissue Protocols, 181-192 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service