Skip to Content
Merck
All Photos(3)

Key Documents

B2133

Sigma-Aldrich

N-Benzoyl-Pro-Phe-Arg-p-nitroanilide hydrochloride

protease substrate, chromogenic, ≥97% (HPLC), powder

Synonym(s):

Bz-Pro-Phe-Arg-p-nitroanilide hydrochloride, Bz-Pro-Phe-Arg-pNA hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H38N8O6 · HCl
CAS Number:
Molecular Weight:
679.17
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

N-Benzoyl-Pro-Phe-Arg-p-nitroanilide hydrochloride, protease substrate

Assay

≥97% (HPLC)

form

powder

solubility

methanol: 25 mg/mL

storage temp.

−20°C

SMILES string

Cl.NC(=N)NCCCC(NC(=O)C(Cc1ccccc1)NC(=O)C2CCCN2C(=O)c3ccccc3)C(=O)Nc4ccc(cc4)N(=O)=O

InChI

1S/C33H38N8O6.ClH/c34-33(35)36-19-7-13-26(29(42)37-24-15-17-25(18-16-24)41(46)47)38-30(43)27(21-22-9-3-1-4-10-22)39-31(44)28-14-8-20-40(28)32(45)23-11-5-2-6-12-23;/h1-6,9-12,15-18,26-28H,7-8,13-14,19-21H2,(H,37,42)(H,38,43)(H,39,44)(H4,34,35,36);1H

InChI key

OGYWLWIIQPDFPW-UHFFFAOYSA-N

Substrates

A chromogenic substrate for Agkistrodon contortrix thrombin-like enzyme and plasma kallikrein.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A V Nieuw Amerongen et al.
Biochimica et biophysica acta, 786(3), 245-251 (1984-05-17)
The previously isolated female submandibular glycoprotein AM1 ( Nieuw Amerongen , A.V., Vreugdenhil , A.P. and Roukema , P.A. (1977) Biochim. Biophys. Acta 495, 324-335) has been shown to have hydrolytic activity using N-alpha-benzoyl-L-arginine ethylester (BAEE) and ChromozymR PK as
G Vogel et al.
Folia haematologica (Leipzig, Germany : 1928), 109(1), 115-120 (1982-01-01)
The kallikrein specific chromogenic peptide substrates S-2302 (KABI) and Chromozym PK (Boehringer) were used in the first analysis of a familial defect in the early stage of clotting. Slight to extensive prolongation of the activated partial thromboplastin time was seen
[Determination of factor XII and plasma prekallikrein with chromozym PK].
J Stürzebecher et al.
Zeitschrift fur medizinische Laboratoriumsdiagnostik, 23(5), 306-311 (1982-10-01)
Determination of factor XII in plasma using the kallikrein substrate chromozym PK.
C Klessen et al.
Thrombosis research, 25(6), 501-505 (1982-03-15)
K Nishikawa et al.
Biochemical pharmacology, 43(6), 1361-1369 (1992-03-17)
Bradykinin (BK), an important mediator of allergic reactions and pain induction, is released by the activation of the plasma kallikrein-kinin (K-K) cascade. Neurotropin is a biological material obtained from inflamed rabbit skin inoculated with vaccinia virus and is widely used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service