Skip to Content
Merck
All Photos(3)

Key Documents

AV54324

Sigma-Aldrich

Anti-IL1B antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-IL-1, Anti-IL1-BETA, Anti-IL1F2, Anti-Interleukin 1, β

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

17 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL1B(3553)

General description

Interleukin-1β (IL1β) is produced by activated macrophages. It is synthesized as pro-IL-1β which is changed to active form IL-1β with the help of pro-inflammatory protease caspase-1. IL-1β belongs to the IL-1 gene family. It is a proinflammatory cytokine. The gene is mapped to human chromosome 2q14.

Immunogen

Synthetic peptide directed towards the N terminal region of human IL1B

Application

Anti-IL1B antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

Interleukins are proinflammatory cytokines produced on cell injury or trauma. There are two closely related types of interleukins produced by the cell, IL-1α and IL-1β that are produced by the activation of NF-κB transcription factor in response to bacterial infection or LPS. Both IL-1α and IL-1β exert their cellular effects by signalling through IL-1 receptor type 1. IL-1β has pleiotropic activities and is protective against infections by recruitment of neutrophils to the infection site, secretion of cytokines and chemokines and activation of adhesion molecules. IL-1β overproduction has been implicated in many auto-immune and allergic disorders such as Cryopirin-associated periodic syndromes, Muckle-Wells syndrome and skin lesions.

Sequence

Synthetic peptide located within the following region: DLDLCPLDGGIQLRISDHHYSKGFRQAASVVVAMDKLRKMLVPCPQTFQE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manoranjan Sahoo et al.
TheScientificWorldJournal, 11, 2037-2050 (2011-11-30)
The inflammasome is an important innate immune pathway that regulates at least two host responses protective against infections: (1) secretion of the proinflammatory cytokines IL-1β and IL-18 and (2) induction of pyroptosis, a form of cell death. Inflammasomes, of which
Charles A Dinarello
Current opinion in pharmacology, 4(4), 378-385 (2004-07-15)
All biological agents currently used for reducing TNFalpha activity in disease are neutralization strategies; however, there are several strategies for reducing interleukin (IL)-1 activities: the IL-1 receptor antagonist (IL-1Ra), anti-IL-1beta monoclonal antibodies, the IL-1 Trap, IL-1 receptor type I antibodies
Axel Weber et al.
Science signaling, 3(105), cm1-cm1 (2010-01-21)
The interleukin-1 (IL-1) family of cytokines comprises 11 proteins (IL-1F1 to IL-1F11) encoded by 11 distinct genes in humans and mice. IL-1-type cytokines are major mediators of innate immune reactions, and blockade of the founding members IL-1alpha or IL-1beta by
Emmanuel Contassot et al.
Swiss medical weekly, 142, w13590-w13590 (2012-06-02)
Interleukin 1, one of the first cytokines discovered in the 1980s, and a potent mediator of fever, pain and inflammation, is at present experiencing a revival in biology and medicine. Whereas the mechanism of activation and secretion of interleukin 1β

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service