Skip to Content
Merck
All Photos(1)

Documents

AV43905

Sigma-Aldrich

Anti-SLC20A1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp686J2397, Anti-FLJ41426, Anti-GLVR1, Anti-Glvr-1, Anti-PIT1, Anti-PiT-1, Anti-Solute carrier family 20 (phosphate transporter), member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

74 kDa

species reactivity

pig, dog, horse, bovine, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC20A1(6574)

Related Categories

General description

Sodium-dependent phosphate transporter 1 (SLC20A1) protein is an inorganic phosphate transporter and a sodium-phosphate symporter. It is expressed in the plasma membrane and belongs to the inorganic phosphate transporter family (PiT). Also called PIT1, it is an N-linked glycosylated protein with N and C termini placed in the extracellular region. It comprises 12 transmembrane domains. SLC20A1 gene is mapped to human chromosome 2q14.1. 

Specificity

Anti-SLC20A1 polyclonal antibody reacts with canine and human solute carrier family 20 (phosphate transporter) member 1 proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of human SLC20A1

Application

Anti-SLC20A1 antibody produced in rabbit has been used in western blotting (1:1000).

Biochem/physiol Actions

Sodium-dependent phosphate transporter 1 (SLC20A1) protein is a high-affinity sodium (Na+)-phosphate (Pi) co-transporter that imports inorganic phosphate (Pi). It serves as a receptor for gibbon ape leukemia virus (GALV), feline leukemia virus type B (FeLV-B), woolly monkey virus and 10A1 murine leukemia virus, making it crucial for viral entry. SLC20A1 is essential for urinary tract and urorectal development. Variants ofSLC20A1 gene are implicated in the pathophysiology of bladder exstrophy-epispadias complex (BEEC) and in cloacal exstrophy. It may also participate in normal growth and development of the liver. Elevated expression of SLC20A1 gene is correlated to the activation of the wingless (Wnt)/β-catenin signaling pathway in somatotroph adenomas. SLC20A1 protein may participate in tumor necrosis factor-induced apoptosis  and regulate cell proliferation, erythroid and B cell differentiation.

Sequence

Synthetic peptide located within the following region: LVALYLVYDTGDVSSKVATPIWLLLYGGVGICVGLWVWGRRVIQTMGKDL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eugénie Koumakis et al.
Scientific reports, 9(1), 1808-1808 (2019-02-14)
PiT1/SLC20A1 is an inorganic phosphate transporter with additional functions including the regulation of TNFα-induced apoptosis, erythropoiesis, cell proliferation and insulin signaling. Recent data suggest a relationship between PiT1 and NF-κB-dependent inflammation: (i) Pit1 mRNA is up-regulated in the context of
Laurent Beck et al.
PloS one, 5(2), e9148-e9148 (2010-02-18)
PiT1 (or SLC20a1) encodes a widely expressed plasma membrane protein functioning as a high-affinity Na(+)-phosphate (Pi) cotransporter. As such, PiT1 is often considered as a ubiquitous supplier of Pi for cellular needs regardless of the lack of experimental data. Although
Mengqi Chang et al.
Frontiers in endocrinology, 11, 596554-596554 (2021-02-13)
Pituitary adenomas (PAs) can be classified as non-secreting adenomas, somatotroph adenomas, corticotroph adenomas, lactotroph adenomas, and thyrotroph adenomas. Substantial advances have been made in our knowledge of the pathobiology of PAs. To obtain a comprehensive understanding of the molecular biological
Karen B Farrell et al.
Journal of virology, 76(15), 7683-7693 (2002-07-05)
The mammalian gammaretroviruses gibbon ape leukemia virus (GALV) and feline leukemia virus subgroup B (FeLV-B) can use the same receptor, Pit1, to infect human cells. A highly polymorphic nine-residue sequence within Pit1, designated region A, has been proposed as the
Johanna Magdalena Rieke et al.
Frontiers in cell and developmental biology, 8, 567-567 (2020-08-28)
Previous studies in developing Xenopus and zebrafish reported that the phosphate transporter slc20a1a is expressed in pronephric kidneys. The recent identification of SLC20A1 as a monoallelic candidate gene for cloacal exstrophy further suggests its involvement in the urinary tract and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service