Skip to Content
Merck
All Photos(3)

Documents

AC-6000

Sigma-Aldrich

HighPrep PCR Clean-up System

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500

form

liquid

packaging

pkg of 250 mL (AC-60250 (HighPrep PCR (250 mL)))
pkg of 5 mL (AC-60005 (HighPrep PCR (5 mL)))
pkg of 50 mL (AC-60050 (HighPrep PCR (50 mL)))
pkg of 500 mL (AC-60500 (HighPrep PCR (500 mL)))

storage condition

do not freeze

storage temp.

2-8°C

General description

Magnetic beads-based reagent for manual and for automated post-PCR and post-enzymatic reaction clean-up, for NGS library clean-up and amplicon size selection.

The HighPrep PCR Clean-up System is a paramagnetic beads-based post PCR clean-up reagent designed for efficient purification of DNA fragments and for size-specific selection of amplicons. The purification consists of removal of salts, primers, primer-dimers, dNTPs, as DNA fragments are selectively bound to the magnetic bead particles. Highly purified DNA is eluted with low salt elution buffer or water which can be used directly for downstream applications, such as library construction, next generation sequencing (NGS), Sanger sequencing, cloning, restriction digestions, adapoter ligation, microarrays, and so on. The magnetic beads are used for different library prep chemistries for NGS.

Application

HighPrep PCR Clean-up System may be used in:
  • Post-PCR and post-enzymatic reaction clean-up
  • NGS library preparation
  • Amplicon size selection for NGS
  • Microarrays
  • PCR
  • Sanger sequencing
  • Restriction enzyme digestions, adapter ligations
  • Cloning

Features and Benefits

  • High recovery of >100 bp amplicons
  • Adaptable to high throughput liquid handling systems
  • Stable recovery of amplicons greater than 100bp, predictable and consistent size selection
  • No centrifugation or filtration leads to higher and purer yields

Storage and Stability

14 months when stored as directed

Legal Information

HiPrep is a trademark of Cytiva
HighPrep is a trademark of MagBio Genomics, Inc.

Disclaimer

For Research Use Only. Not for Use in Diagnostic Procedures.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soil bacterial diversity under conservation agriculture-based cereal systems in Indo-Gangetic Plains
Choudhary M, et al.
3 Biotech, 8, 304-304 (2018)
Signature of genome wide gene expression in classical swine fever virus infected macrophages and PBMCs of indigenous vis-a-vis crossbred pigs
Sah V, et al
Gene, 144356-144356 (2020)
N Salvetat et al.
Journal of neurovirology, 25(6), 825-836 (2019-07-25)
Treatment-emergent depression is a common complication in patients with chronic hepatitis C virus (HCV) infection undergoing antiviral combination therapy with IFN-α and ribavirin. It has recently been shown that changes in A-to-I RNA editing rates are associated with various pathologies
Vaishali Sah et al.
Gene, 731, 144356-144356 (2020-01-15)
The genetic basis of differential host immune response vis-à-vis transcriptome profile was explored in PBMCs of indigenous (Ghurrah) and crossbred pigs after classical swine fever vaccination and in monocyte derived macrophages (MDMs) challenged with virulent classical swine fever (CSF) virus.
Deep sequencing of near full-length HIV-1 genomes from plasma identifies circulating subtype C and infrequent occurrence of AC recombinant form in Southern India}
Alampalli SV, et al.
PLoS ONE, 17 (2017)

Protocols

HighPrep™ PCR purification magnetic clean-up beads for manual and automated post-PCR, post-enzymatic reaction, NGS library clean-up.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service