Skip to Content
Merck
All Photos(1)

Key Documents

A9730

Sigma-Aldrich

Adefovir dipivoxil

Synonym(s):

9-(2[bis(Pivaloyloxymethoxy)phosphorylmethoxy]ethyl)adenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32N5O8P
CAS Number:
Molecular Weight:
501.47
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.76

biological source

synthetic

Assay

≥98% (HPLC)

form

solid

solubility

ethanol: 50 mg/mL

antibiotic activity spectrum

viruses

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

CC(C)(C)C(=O)OCOP(=O)(COCCn1cnc2c(N)ncnc12)OCOC(=O)C(C)(C)C

InChI

1S/C20H32N5O8P/c1-19(2,3)17(26)30-11-32-34(28,33-12-31-18(27)20(4,5)6)13-29-8-7-25-10-24-14-15(21)22-9-23-16(14)25/h9-10H,7-8,11-13H2,1-6H3,(H2,21,22,23)

InChI key

WOZSCQDILHKSGG-UHFFFAOYSA-N

Biochem/physiol Actions

Adefovir is an antiviral acyclic nucleoside phosphonate (ANP) analog that works by blocking reverse transcriptase. It is an inhibitor of duck hepatitis B virus (DHBV) replication that inhibits covalently closed circular DNA (CCC DNA) amplification. Cytotxicity is induced by Human Renal Organic Anion Transporter 1 (hOAT1). Adefovir is an Inhibitor of edema factor (EF)-induced cAMP accumulation and changes in cytokine production in mouse primary macrophages .

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick Marcellin et al.
The New England journal of medicine, 348(9), 808-816 (2003-02-28)
In preclinical and phase 2 studies, adefovir dipivoxil demonstrated potent activity against hepatitis B virus (HBV), including lamivudine-resistant strains. We randomly assigned 515 patients with chronic hepatitis B who were positive for hepatitis B e antigen (HBeAg) to receive 10
Man-Fung Yuen et al.
Expert opinion on pharmacotherapy, 5(11), 2361-2367 (2004-10-27)
Adefovir dipivoxil, an acyclic nucleotide analogue, is effective for the treatment of chronic hepatitis B in both hepatitis B e antigen (HBeAg)-positive and -negative patients, with improvement in liver histology, hepatitis B virus (HBV) DNA levels, alanine aminotransferase levels, and
Myron J Tong et al.
Seminars in liver disease, 24 Suppl 1, 37-44 (2004-06-12)
Adefovir dipivoxil, a nucleotide analog of adenosine monophosphate, is an antiviral agent that suppresses hepatitis B virus (HBV) replication through inhibition of DNA polymerase and by chain termination. To determine the effectiveness of adefovir, three populations of patients with chronic
J Jones et al.
Health technology assessment (Winchester, England), 13(35), 1-172 (2009-07-18)
To update and extend a 2006 report on the clinical effectiveness and cost-effectiveness of adefovir dipivoxil (ADV) and pegylated interferon alpha (PEG-alpha) for the treatment of chronic hepatitis B (CHB). Thirteen bibliographic databases were searched including MEDLINE, EMBASE and the
Xiaokun Shen et al.
Scientific reports, 6, 38778-38778 (2016-12-13)
A pressing need exists for improved therapeutic options for chronic hepatitis B (CHB). Pegylated-interferon-alpha (Peg-IFN-α) achieves sustained off-treatment responses in many cases because of its direct anti-viral effects and regulation of the immune response. However, non-responsiveness to Peg-IFN-α is frequent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service