Skip to Content
Merck
All Photos(2)

Documents

A9546

Sigma-Aldrich

2-Amino-6-methylmercaptopurine

≥95%

Synonym(s):

2-Amino-6-methylthiopurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N5S
CAS Number:
Molecular Weight:
181.22
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥95%

form

solid

SMILES string

[H]n1cnc2c(SC)nc(N)nc12

InChI

1S/C6H7N5S/c1-12-5-3-4(9-2-8-3)10-6(7)11-5/h2H,1H3,(H3,7,8,9,10,11)

InChI key

YEGKYFQLKYGHAR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Amino-6-methylmercaptopurine (6-MTG) has been used as a supplement in Dulbecco′s modified Eagles medium (DMEM) medium for the selection of gpt-expressing recombinant virus mCMVhMIEPE-gpt.lacZ (cytomegalovirus major immediate-early promotor-enhancer complex-gpt.lacz). It has also been used as a standard in high performance liquid chromatography (HPLC) to assess the activity of thiopurine methyltransferase (TPMT) enzyme.
2-Amino-6-methylmercaptopurine is a 2-amino-6-alkyldithiopurine that has been used with other 6-position carbon analogues to study brain specific diazepam binding.

Biochem/physiol Actions

2-Amino-6-methylmercaptopurine is synthesized from 6-mercaptopurine by the S methylation activity of thiopurine methyl transferase (TMPT) enzyme.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

T Dooley et al.
Journal of chromatography, 337(2), 321-327 (1985-02-08)
A flow-fluorimetric high-performance liquid chromatographic assay for 6-methylthioguanine in urine has been developed. This compound is a major catabolite of 6-thioguanine, an important drug in cancer chemotherapy. The metabolite was extracted from alkaline urine with ethyl acetate which was injected
T R Waters et al.
Biochemistry, 36(9), 2501-2506 (1997-03-04)
It has been suggested that the cytotoxicity of 6-thioguanine depends upon (1) incorporation of 6-thioguanine into DNA, (2) methylation by S-adenosylmethionine (SAM) of the thio group to give S6-methylthioguanine, (3) miscoding during DNA replication to give [SMeG] x T base
Cancer Chemotherapy and Biotherapy: Principles and Practice, 193-193 (2010)
Bifeng Yuan et al.
ACS chemical biology, 5(11), 1021-1027 (2010-09-03)
Thiopurines are effective immunosuppressants and anticancer agents. However, the long-term use of thiopurines was found to be associated with a significantly increased risk of various types of cancer. To date, the specific mechanism(s) underlying the carcinogenicity associated with thiopurine treatment
Hongxia Wang et al.
Analytical chemistry, 82(13), 5797-5803 (2010-06-17)
Thiopurines, including mercaptopurine (MP), 6-thioguanine ((S)G) and azathioprine, are widely used for the treatment of many human diseases including acute lymphoblastic leukemia (ALL). To exert their cytotoxic effect, these prodrugs need to be metabolically activated to (S)G nucleotides and incorporated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service