Skip to Content
Merck
All Photos(1)

Documents

A8967

Sigma-Aldrich

Anti-Amyloid Precursor Protein, N-Terminal antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-APP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 95-100 kDa

species reactivity

human, rat, mouse

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200 using formic acid-treated sections of human Alzheimer′s disease (AD) brain
microarray: suitable
western blot: 1:1,000 using rat brain extract or supernatant of 293T cells secreting APP.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APP(351)
mouse ... App(11820)
rat ... App(54226)

General description

Amyloid precursor proteins (APPs) are transmembrane glycoproteins that are found in a wide range of tissues. APPs have 3 main isoforms, namely, APP695, APP751 and APP770 that are derived from alternative splicing events in cells. It is expressed at high levels in the brain. APP gene is mapped to human chromosome 21q11.2-q21. It is a 695 amino acid protein which possesses a large ectodomain and comparatively short intracellular region.
The immunogen sequence is identical to the APP isoforms APP751 and APP770 and is highly conserved (single amino acid substitution) in rat and mouse APP695. The antibody recognizes APP695, APP751 and APP770.

Immunogen

synthetic peptide corresponding to the N-terminal of human APP695 (amino acids 46-60) conjugated to KLH.

Application

Anti-Amyloid Precursor Protein, N-Terminal antibody produced in rabbit has been used in:
  • western blotting
  • immunostaining
  • immunofluorescence

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
SH-SY5Y cell lysates were analyzed by western blot using rabbit anti-Amyloid Precursor Protein, C-Terminal as the primary antibody at a 1:500 dilution.

Biochem/physiol Actions

Amyloid precursor proteins (APPs) regulates cell growth, motility, neurite outgrowth and cell survival. The intracellular C-terminus of APP serves as a transcriptional regulator and as a receptor for kinesin-1-mediated axonal transport. Alzheimer′s disease is characterized by deposition of amyloid in the central nervous system, in neurite plaques and on cerebral vasculature. Mutations in the APP gene are linked with rare forms of autosomal dominant familial Alzheimer′s Disease (FAD). APPs undergo post-translational processing including N- and O-glycosylation, phosphorylation and sulfation.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Association of TrkA and APP Is Promoted by NGF and Reduced by Cell Death-Promoting Agents
Canu N, et al.
Frontiers in Molecular Neuroscience, 10 (2017)
Dopamine induces apoptosis in APPswe-expressing Neuro2A cells following Pepstatin-sensitive proteolysis of APP in acid compartments
Cagnin M, et al.
Brain Research, 1471, 102-117 (2012)
Homo-and Heterodimerization of Proteins in Cell Signaling: Inhibition and Drug Design
Advances in Protein Chemistry and Structural Biology, 111, 1-59 (2018)
The Alzheimer amyloid precursor protein maps to human chromosome 21 bands q21. 105-q21. 05
Korenberg JR, et al.
Genomics, 5(1), 124-127 (1989)
Migration of blood cells to beta-amyloid plaques in Alzheimer's disease
Hohsfield LA, et al.
Experimental Gerontology, 65, 8-15 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service