Skip to Content
Merck
All Photos(1)

Documents

A8580

Sigma-Aldrich

Anti-Glutathione-S-Transferase (GST)–Agarose antibody produced in rabbit

IgG fraction of antiserum, PBS suspension

Synonym(s):

Anti-Glutathione-S-Transferase, Anti-GST

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

agarose conjugate

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

PBS suspension

mol wt

antigen 27.5 kDa

should not react with

porcine, bovine, human, rabbit, rat

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... GSTP1(2950)

Looking for similar products? Visit Product Comparison Guide

General description

Glutathione-S-transferases (GSTs) are dimeric proteins, which belongs to phase II detoxification enzymes family. The highly polymorphic human cytosolic GSTs are divided into six classes, such as, α, μ, ω, π, θ, and ζ.
The antibody is specific for native as well as denatured-reduced forms of glutathione-S-transferase from Schistosoma japonicum. Anti-GST may be used in various immunoassays to identify the expression of GST fusion proteins.

Immunogen

recombinant GST from Schistosoma japonicum expressed in E. coli.

Application

Immunoprecipitation was performed on NIH 3T3 cells transiently transfected with GST fusion proteins. IP was performed using agarose-linked rabbit anti-GST IgG with mixing for 2 hours at 4 degrees.
Rabbit Anti-Glutathione-S-Transferase (GST)-Agarose antibody can be used for ELISA, immunoaffinity purification and immunoprecipitation assays.

Biochem/physiol Actions

Glutathione-S-transferases (GSTs) can catalyse the conjugation of glutathione (GSH) to various endogenous and exogenous electrophilic compounds. The π and μ classes of GSTs controls the mitogen-activated protein (MAP) kinase pathway. It can also serve as transport proteins.

Physical form

Suspension in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Preparation Note

Prepared by coupling cyanogen bromide-activated agarose at 7-8 mg antibody per mL resin volume.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolism of xenobiotics of human environments
Progress in Molecular Biology and Translational Science, 112, 31-88 (2012)
Clinical pathology in non-clinical toxicology testing
Haschek and Rousseaux's Handbook of Toxicologic Pathology, 112(47), 565-594 (2013)
Xun Shang et al.
The Journal of biological chemistry, 282(12), 8801-8811 (2007-02-03)
p200 RhoGAP, a member of the Rho GTPase-activating protein (RhoGAP) family, was previously implicated in the regulation of neurite outgrowth through its RhoGAP activity. Here we show that ectopic expression of p200 RhoGAP stimulates fibroblast cell proliferation and cell cycle
Bin Sheng et al.
Cell death & disease, 12(11), 1074-1074 (2021-11-12)
Deubiquitinases (DUBs) have important biological functions, but their roles in breast cancer metastasis are not completely clear. In this study, through screening a series of DUBs related to breast cancer distant metastasis-free survival (DMFS) in the Kaplan-Meier Plotter database, we
The role of glutathione-S-transferase in anti-cancer drug resistance
Townsend D M and Tew K D
Oncogene, 22(47), 7369-7369 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service