Skip to Content
Merck
All Photos(2)

Key Documents

A7607

Sigma-Aldrich

Anti-α-Smooth Muscle Actin (ACTA2) Antibody,

mouse monoclonal, CGA7

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Monoclonal Anti-Actin, Smooth Muscle antibody produced in mouse, ~1.0 mg/mL, clone CGA7, purified immunoglobulin, buffered aqueous solution

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CGA7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~42 kDa

species reactivity

rat, chicken, mouse, human, monkey

concentration

~1.0 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: 5-10 μg/mL using chicken gizzard total extract

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

chicken ... ACTA2(423787)
human ... ACTA2(59)
mouse ... Acta2(11475)
rat ... Acta2(81633)

General description

Actin is a highly conserved protein that is a major component of both the cytoskeletal and contractile structures in all cell types. Its expression is related to the type of differentiation and to the functional state of cells and tissues. Actin can be found in two different forms of aggregation, the globular or the fibrillar state, and at least six distinct isoforms occur in vertebrates. The actins exhibit over 90% sequence homology, but each isoform has a unique NH2-terminal sequence. The isoforms are comprised of three alpha actins (skeletal, cardiac, smooth), one beta actin (beta-non-muscle) and two gamma actins (gamma smooth muscle and gamma non-muscle).
Monoclonal Anti-Actin, Smooth Muscle, reacts with human, monkey, chicken, rat, and mouse smooth muscle actin.
Monoclonal Anti-Actin, Smooth Muscle (mouse IgG2b isotype) is derived from the hybridoma CGA7 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with partially purified chicken gizzard actin. α-actin 2 (ACTA2) codes for a smooth muscle specific α-actin.
The antibody specifically recognizes α and γ smooth muscle actin.

Immunogen

partially purified chicken gizzard actin.

Application

Monoclonal Anti-Actin, Smooth Muscle antibody produced in mouse has been used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • immunoblotting
  • immunocytochemistry
  • immunohistochemistry

Biochem/physiol Actions

Actin and myosin are constituents of many cell types and are involved in a myriad of cellular processes including locomotion, secretion, cytoplasmic streaming, phagocytosis and cytokinesis. Mutations in alpha-actin 2 (ACTA2) results in thoracic aortic aneurysm, Moyamoya disease, coronary artery disease, stroke and multisystemic smooth muscle dysfunction syndrome. It plays a functional role in the smooth muscle of non-vascular tissues.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beneficial effects of galectin-3 blockade in vascular and aortic valve alterations in an experimental pressure overload model
Ibarrola J, et al.
International Journal of Molecular Sciences, 18(8), 1664-1664 (2017)
M Rodriguez et al.
Oncogene, 35(9), 1153-1162 (2015-06-02)
Among frequent events in prostate cancer are loss of the tumor-suppressor phosphatase and tensin homologue (PTEN) and overexpression of prostate-specific G-protein-coupled receptor (PSGR), but the potential tumorigenic synergy between these lesions is unknown. Here, we report a new mouse model
Comparison of human olfactory and skeletal MSCs using osteogenic nanotopography to demonstrate bone-specific bioactivity of the surfaces
Johnstone SA, et al.
Acta Biomaterialia, 13, 266-276 (2015)
ACTA2 mutation and postpartum hemorrhage: a case report
Cooper K and Brown S
BMC Medical Genetics, 18(1), 143-143 (2017)
Role of fibroblast growth factor signaling during proepicardium formation in the chick embryo
Torlopp A, et al.
Developmental Dynamics, 239(9), 2393-2403 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service