Skip to Content
Merck
All Photos(3)

Key Documents

A7562

Sigma-Aldrich

Acetobromo-α-D-galactose

≥93% (TLC)

Synonym(s):

2,3,4,6-Tetra-O-acetyl-α-D-galactopyranosyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19BrO9
CAS Number:
Molecular Weight:
411.20
Beilstein:
96671
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥93% (TLC)

form

powder

optical activity

[α]25/D 200 to 225 °, c = 0.5% (w/v) in chloroform

contains

1% CaCO3 as stabilizer

color

white to off-white

solubility

chloroform: soluble 5 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@H]1OC(C)=O

InChI

1S/C14H19BrO9/c1-6(16)20-5-10-11(21-7(2)17)12(22-8(3)18)13(14(15)24-10)23-9(4)19/h10-14H,5H2,1-4H3/t10-,11+,12+,13-,14+/m1/s1

InChI key

CYAYKKUWALRRPA-HTOAHKCRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,3,4,6-Tetra-O-acetyl-α-D-galactopyranosyl bromide is used as an organic condensation reagent for chemistries such as N- and S- galactosylation reactions and the synthesis of spacer-equipped phosphorylated di-, tri- and tetrasaccharide fragments of the O-specific polysaccharides.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdel Kader Mansour et al.
Nucleosides, nucleotides & nucleic acids, 22(9), 1825-1833 (2003-10-10)
The 1-(2,3,4,6-tetra-O-acetyl-beta-D-galactopyranosyl)-3-aryl-5-benzyl (or substituted benzyl)-1,2,4-triazin-6(1H)-/ones or thiones were prepared via galactosidation of 3-aryl-5-benzyl (or substituted benzyl)-1,2,4-triazin-6(1H)-/ones or thiones with 2,3,4,6-tetra-O-acetyl-alpha-D-galactopyranosyl bromide. The structure of the new galactosyl derivatives was based on both spectroscopic and chemical evidences.
Jiajia Qian et al.
The Journal of pharmacy and pharmacology, 71(6), 929-936 (2019-03-06)
Resveratrol (Res), a naturally occurring polyphenol, has shown pharmacological activities in treatment of liver diseases. However, the application of Res was limited by its poor bioavailability and liver targeting. Herein, 3-O-β-D-Galactosylated Resveratrol (Gal-Res) was synthesized by structural modification of Res
Bart Ruttens et al.
Carbohydrate research, 341(9), 1077-1080 (2006-05-03)
The synthesis of oligosaccharide fragments of the O-specific polysaccharide of Vibrio cholerae O139 containing a 4,6-cyclic phosphate galactose residue linked to GlcNAc is described. 8-Azido-3,6-dioxaoctyl 2,3,4,6-tetra-O-acetyl-beta-D-galactopyranosyl-(1-->3)-2-acetamido-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranoside, obtained by condensation of 2,3,4,6-tetra-O-acetyl-alpha-D-galactopyranosyl bromide and 8-azido-3,6-dioxaoctyl 2-acetamido-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranoside, was converted to 8-azido-3,6-dioxaoctyl 3-O-benzyl-beta-D-galactopyranosyl-(1-->3)-2-acetamido-6-O-benzyl-2-deoxy-beta-D-glucopyranoside
David J Claffey et al.
Carbohydrate research, 339(14), 2433-2440 (2004-09-25)
Condensation of 2,3,4,6-tetra-O-acetyl-alpha-D-glucopyranosyl-, 2,3,4-tri-O-acetyl-alpha-D-xylopyranosyl- and of 2,3,4,6-tetra-O-acetyl-alpha-D-galactopyranosyl bromides with l,4:3,6-dianhydro-D-glucitol under Koenigs-Knorr conditions, and using the Helferich modification of the reaction showed regioselectivity in glysosylation at C-5 of isosorbide.
Ahmed I Khodair et al.
Carbohydrate research, 346(18), 2831-2837 (2011-11-18)
N- and S-galactosylation was carried out via the reaction of 5-((Z)-arylidene)-2-thioxo-4-thiazolidinones with 2,3,4,6-tetra-O-acetyl-α-d-galactopyranosyl bromide under alkaline conditions or under silylation conditions. Deacetylation of the N-galactosylation products was performed with concentrated hydrochloric acid in methanol (3.5%) or sodium methoxide in methanol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service