A5752
Adrenochrome
>95% purity (HPLC), powder
Synonym(s):
3-Hydroxy-1-methyl-5,6-indolinedione
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Empirical Formula (Hill Notation):
C9H9NO3
CAS Number:
Molecular Weight:
179.17
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Recommended Products
Product Name
Adrenochrome, ≥95% (HPLC)
Assay
≥95% (HPLC)
form
powder
color
red to brown
εmax
3-4.5 at 485-490 nm in water
9.0-10.5 at 299-304 nm in water
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
−20°C
SMILES string
CN1CC(O)C2=CC(=O)C(=O)C=C12
InChI
1S/C9H9NO3/c1-10-4-9(13)5-2-7(11)8(12)3-6(5)10/h2-3,9,13H,4H2,1H3
InChI key
RPHLQSHHTJORHI-UHFFFAOYSA-N
Related Categories
Application
Adrenochrome has been used to study the effect of adrenochrome on coronary artery constriction in the rat heart.
Biochem/physiol Actions
Adrenochrome is an oxidation product of adrenaline. It has been implicated in cardiotoxicity. It is a short-lived cytotoxic molecule and is part of the leukocyte response used to kill bacteria. Adrenochrome is also suggested to exhibit psychotomimetic effects. It is used to determine superoxide dismutase activity.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Janna Shainsky et al.
Applied and environmental microbiology, 75(14), 4711-4719 (2009-05-26)
Enantiopure sulfoxides are prevalent in drugs and are useful chiral auxiliaries in organic synthesis. The biocatalytic enantioselective oxidation of prochiral sulfides is a direct and economical approach for the synthesis of optically pure sulfoxides. The selection of suitable biocatalysts requires
Vera Marisa Costa et al.
Chemical research in toxicology, 20(8), 1183-1191 (2007-07-17)
High concentrations of circulating biogenic catecholamines often exist during the course of several cardiovascular disorders. Additionally, coronary dysfunctions are prominent and frequently related to the ischemic and reperfusion phenomenon (I/R) in the heart, which leads to the release of large
The oxidative metabolism of catecholamines in the brain: a review.
Smythies J and Galzigna L
Biochimica et Biophysica Acta, 1380, 159-159 (1998)
The adrenochrome hypothesis of schizophrenia revisited.
Smythies J
Neurotoxicity Research, 4, 147-147 (2002)
Ventricular dysfunction and necrosis produced by adrenochrome metabolite of epinephrine: relation to pathogenesis of catecholamine cardiomyopathy.
Yates JC, et al.
American Heart Journal, 102, 210-210 (1981)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service