A4979
Anti-Water Channel Aquaporin 5 antibody produced in rabbit
affinity isolated antibody, lyophilized powder
Synonym(s):
Anti-AQP-5, Anti-PPKB
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
lyophilized powder
species reactivity
mouse, rat
technique(s)
western blot: 1:200-1:500 using mouse salivary gland and rat lung
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
mouse ... Aqp5(11830)
rat ... Aqp5(25241)
General description
Anti-Water Channel Aquaporin 5 binds to aquaporin 5 from mouse salivary gland and rat lung membranes.
Aquaporin-5 is part of a family of aquaporins that function to transport water across cell membranes. AQP5 is involved in tear, saliva, and sweat production.
Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0).
Immunogen
peptide corresponding to residues 251-265 of rat AQP5. The sequence is identical in mouse and 10/16 residues identical in human.
Application
Rabbit Anti-Water Channel Aquaporin 5 antibody can be used for western blot at a dilution of 1:200-1:500 using mouse salivary gland and rat lung tissue extracts.
Physical form
Lyophilized from phosphate buffered saline, pH 7.4, 1% BSA, ≤0.1% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J W Snuggs et al.
Scientific reports, 11(1), 3164-3164 (2021-02-06)
The central region of the intervertebral disc (IVD) is rich in proteoglycans, leading to a hyperosmotic environment, which fluctuates with daily loading. The cells of the nucleus pulposus (NP cells) have adapted to this environment via the function of tonicity
Michael G Haase et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 62(5), 355-368 (2014-03-29)
Ionizing radiation (IR) leads to fibrosing alveolitis (FA) after a lag period of several weeks to months. In a rat model, FA starts at 8 weeks after IR. Before that, at 5.5 weeks after IR, the transcription factors Sp1 (stimulating
Mulugu V Brahmajothi et al.
Antioxidants & redox signaling, 21(13), 1823-1836 (2014-08-05)
The aims of this study were to determine hyperoxia effects on S-nitrosothiol (SNO) accumulation and L-type amino acid transporter 1 (LAT1) expression/function in alveolar epithelium and to determine whether hyperoxia impairs exogenous nitric oxide (NO) treatment effects in alveolar epithelium
Hongyu Wu et al.
PloS one, 8(1), e53342-e53342 (2013-01-18)
Dot1l encodes histone H3 K79 methyltransferase Dot1a. Mice with Dot1l deficiency in renal Aqp2-expressing cells (Dot1l(AC)) develop polyuria by unknown mechanisms. Here, we report that Aqp5 links Dot1l deletion to polyuria through Aqp2. cDNA array analysis revealed and real-time RT-qPCR
Christina Vohlen et al.
Cells, 10(11) (2021-11-28)
Prematurely born infants often require supplemental oxygen that impairs lung growth and results in arrest of alveolarization and bronchopulmonary dysplasia (BPD). The growth hormone (GH)- and insulin-like growth factor (IGF)1 systems regulate cell homeostasis and organ development. Since IGF1 is
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service