Skip to Content
Merck
All Photos(5)

Key Documents

A4393

Sigma-Aldrich

R-(−)-Apomorphine hydrochloride hemihydrate

calcined, ≥98% (with NaOH, titration)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C17H17NO2 · HCl · 1/2H2O
CAS Number:
Molecular Weight:
312.79
EC Number:
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (with NaOH, titration)

form

calcined
(Powder to Crystalline Powder)

storage condition

protect from light
under inert gas

color

white to gray

mp

285-287 °C (lit.)

solubility

H2O: ~10 mg/mL, clear, yellow-green

SMILES string

Cl[H].Cl[H].[H]O[H].[H][C@]12Cc3ccc(O)c(O)c3-c4cccc(CCN1C)c24.[H][C@]56Cc7ccc(O)c(O)c7-c8cccc(CCN5C)c68

InChI

1S/2C17H17NO2.2ClH.H2O/c2*1-18-8-7-10-3-2-4-12-15(10)13(18)9-11-5-6-14(19)17(20)16(11)12;;;/h2*2-6,13,19-20H,7-9H2,1H3;2*1H;1H2/t2*13-;;;/m11.../s1

InChI key

CXWQXGNFZLHLHQ-DPFCLETOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

R-(-)-Apomorphine hydrochloride hemihydrate has been used as a dopamine receptor agonist to study its effects of rotational behavior in rat models of Parkinson′s disease.

Biochem/physiol Actions

R-(−)-Apomorphine acts as a non-selective D1 and D2 dopamine receptor agonist. It shows therapeutic effects against Parkinson′s disease and male erectile dysfunction. R-(−)-Apomorphine also shows neuroprotective, radical scavenging, and emetic effects. Apomorphine stimulates the brain chemoreceptor trigger zone. It stimulates the chemoreceptor trigger zone in the brain.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Li et al.
Neuron, 76(5), 1030-1041 (2012-12-12)
Much recent discussion about the origin of Parkinsonian symptoms has centered around the idea that they arise with the increase of beta frequency waves in the EEG. This activity may be closely related to an oscillation between subthalamic nucleus (STN)
Y H Dang et al.
Neuroscience, 169(4), 1872-1880 (2010-07-06)
The present study examined the role of dopamine and D(1)-and D(2)-like dopamine receptors in ventrolateral orbital cortex (VLO)-evoked anti-hypersensitivity in a rat model of neuropathic pain, as well as the possible underlying mechanisms. Results showed that microinjection of apomorphine [(R(-)-apomorphine
Kristina Malinovskaja et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 83(3), 477-484 (2012-12-05)
The objective of this study was to test a drug delivery system that combines iontophoresis and cation-exchange fibers as drug matrices for the controlled transdermal delivery of antiparkinsonian drug apomorphine. Positively charged apomorphine was bound to the ion-exchange groups of
Fumitoshi Kodaka et al.
European journal of nuclear medicine and molecular imaging, 40(4), 574-579 (2012-12-15)
Dopamine D receptors (DRs) have two affinity states for endogenous dopamine, referred to as high-affinity state (D ), which has a high affinity for endogenous dopamine, and low-affinity state (D ). The density of D can be measured with (R)-2-CHO-N-n-propylnorapomorphine
Anna Sancho-Balsells et al.
Frontiers in cellular neuroscience, 14, 93-93 (2020-06-02)
The role of the WDFY1 protein has been studied as a TLR3/4 scaffold/recruiting protein in the immune system and in different oncogenic conditions. However, its function in brain remains poorly understood. We have found that in mice devoid of Helios

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service