Skip to Content
Merck
All Photos(1)

Documents

A3128

Sigma-Aldrich

Ala-Phe

≥98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16N2O3
CAS Number:
Molecular Weight:
236.27
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

product name

Ala-Phe,

Assay

≥98% (TLC)

Quality Level

form

powder

color

white to off-white

storage temp.

−20°C

SMILES string

C[C@H](N)C(=O)N[C@@H](Cc1ccccc1)C(O)=O

InChI

1S/C12H16N2O3/c1-8(13)11(15)14-10(12(16)17)7-9-5-3-2-4-6-9/h2-6,8,10H,7,13H2,1H3,(H,14,15)(H,16,17)/t8-,10-/m0/s1

InChI key

OMNVYXHOSHNURL-WPRPVWTQSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Ala-Phe

Biochem/physiol Actions

Alanyl dipeptides such as ala-leu, ala-lys, ala-gly, ala-pro, ala-tyr and ala-phe may be used in physicochemical studies or to evaluate dipeptide separation technologies. Alanyl dipeptides may also be used for studying cell uptake mechanisms, dipeptide metabolism or cell growth supplementation benefits.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Lin Zhu et al.
Journal of agricultural and food chemistry, 55(21), 8367-8371 (2007-10-12)
A replicate heart-cut column-switching HPLC method combined with two switching valves was newly developed for the simultaneous determination of three antihypertensive peptides (Ala-Phe, Tyr-Pro, and Trp-Tyr) in tryptic casein hydrolysate in one run-in assay. After a first separation on an
Kyung Min Choi et al.
Analytical biochemistry, 367(2), 167-172 (2007-06-30)
A spectrophotometric assay to determine peptide transport has been developed. Using two chromogenic peptide mimetics, L-phenylalanyl-L-2-sulfanilylglycine (PSG) and L-phenylalanyl-L-3-thiaphenylalanine (PSP), the peptide transport patterns in individual cell species can be evaluated effectively. After the addition of PSG to a HeLa
J Li et al.
Electrophoresis, 20(1), 171-179 (1999-03-05)
The separation of stereoisomers, particularly enantiomers, is important when their physiological activity differs. We have resolved the four stereoisomers each of alanylphenylalanine (Ala-Phe) and of leucylphenylalanine (Leu-Phe) by capillary electrophoresis using beta-cyclodextrin as a buffer additive and urea to enhance
Zhi-Xia Zheng et al.
Electrophoresis, 26(4-5), 1007-1012 (2005-02-17)
A ligand-exchange capillary electrophoresis was explored, with L-ornithine as the ligand and copper(II) as the central ion. Its applicability was demonstrated with underivatized and dansyl amino acids, a dipeptide, and drugs with amino alcohol structure. The enantioselectivity was found to
Yasuhito Shomura et al.
Protein science : a publication of the Protein Society, 21(5), 707-716 (2012-03-13)
BacD is an ATP-dependent dipeptide ligase responsible for the biosynthesis of L-alanyl-L-anticapsin, a precursor of an antibiotic produced by Bacillus spp. In contrast to the well-studied and phylogenetically related D-alanine: D-alanine ligase (Ddl), BacD synthesizes dipeptides using L-amino acids as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service