Skip to Content
Merck
All Photos(2)

Key Documents

A2050

Sigma-Aldrich

Avidin–FITC from egg white

buffered aqueous solution

Synonym(s):

Avidin–Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

conjugate

FITC conjugate

Quality Level

form

buffered aqueous solution

extent of labeling

3-5.5 mol fluorochrome per mol protein

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 20-40 μg/mL
indirect immunofluorescence: 1:200

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

chicken ... AVD(396260)

Looking for similar products? Visit Product Comparison Guide

General description

Avidin is homotetrameric protein (68 kDa) that is obtained from egg whites.

Application

Avidin-FITC from egg white has been used in:
  • flow cytometry
  • double immunostaining
  • double immunofluorescence
  • immunnohistochemistry

To determine the amount of labeled polyclonal IgG present in spores in an IgG prep. A known concentration of FITC avidin from egg white and biotin-IgG avidin was used in excess at 300nm.

Physical form

Solution in 0.01 M phosphate buffered saline containing 10% (v/v) 0.5 M carbonate buffer, pH 9.5, and 15 mM sodium azide.

Preparation Note

Affinity purified protein.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Anthony Y Huang et al.
British journal of pharmacology, 175(7), 1039-1053 (2018-01-13)
Capsaicin-mediated modulation of taste nerve responses is thought to be produced indirectly by the actions of neuropeptides, for example, CGRP and substance P (SP), on taste cells implying they play a role in taste sensitivity. During the processing of gustatory
Jinjie Liu et al.
Experimental and therapeutic medicine, 19(3), 2103-2112 (2020-02-28)
Numerous cases of spinal cord injury following seizure have been previously reported. However, whether spinal cord injury is a common occurrence after seizures and its underlying mechanisms remain unclear. The present study generated a Sprague-Dawley rat model of temporal lobe
Linlin Sun et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(33), 6595-6607 (2019-06-12)
Expressional changes of pain-associated genes in primary sensory neurons of DRG are critical for neuropathic pain genesis. DNA methyltransferase (DNMT)-triggered DNA methylation silences gene expression. We show here that DNMT1, a canonical maintenance methyltransferase, acts as the de novo DNMT
Stephanie G Grancharov et al.
The journal of physical chemistry. B, 109(26), 13030-13035 (2006-07-21)
Monodisperse magnetic nanoparticles (NPs) could enable the ultra-sensitive magnetic detection of biological analytes. However, rendering these particles biocompatible has remained a challenge. We report the bio-functionalization and detection of 12-nm manganese ferrite NPs. We have achieved the site-specific binding of
Substance P as a putative efferent transmitter mediates GABAergic inhibition in mouse taste buds
Huang AY and Wu SY
British Journal of Pharmacology, 175(7), 1039-1053 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service