Skip to Content
Merck

A1900

Albumin, Monomer bovine

Monomer ≥97 %

Synonym(s):

Albumin powder, Bovine albumins

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
eCl@ss:
42010201
EC Number:
232-936-2
NACRES:
NA.26
MDL number:
Biological source:
bovine
Assay:
≥97% (PAGE)
Form:
lyophilized powder
Technique(s):
SDS-PAGE: suitable
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Segment

assay

≥97% (PAGE)

form

lyophilized powder

composition

Monomer, ≥97%

technique(s)

SDS-PAGE: suitable

solubility

H2O: soluble 50 mg/mL

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

General description

The most abundant plasma protein in mammals is albumin. It is produced in the liver and transferred as a non-glycosylated protein into the plasma. This multifunctional protein can bind to ligands.

Application

Albumin, Monomer bovine has been used as a:
  • molecular weight (MW) and polydispersion index (PDI) reference standard for the analysis of hyaluronic acid (HA)-based eye drops
  • standard in multi-angle light scattering coupled with size exclusion chromatography (SEC-MALS) for calibration
  • model protein to generate subvisible particles (SVPs) for flow imaging microscopy studies

The binding properties of albumin can be used for a wide range of clinical, pharmaceutical, and biochemical applications.

Biochem/physiol Actions

Albumin is the most important circulatory protein involved in the control of Ca2+ (and Mg2+) levels in mammals.

Preparation Note

Isolated from albumin (A4503) as described for mercaptalbumin in Janatova, J., et al., J. Biol. Chem., 243, 3612 (1968).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library