Skip to Content
Merck
All Photos(1)

Key Documents

A0851

Sigma-Aldrich

Anti-Angiopoietin-2 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Ang-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
IHC (p)
WB
clone:
polyclonal
species reactivity:
human
citations:
3
technique(s):
ELISA: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 5-15 μg/mL using human tissue
western blot: 0.1-0.2 μg/mL

biological source

goat

Quality Level

recombinant

expressed in goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

antigen ~66 kDa

species reactivity

human

technique(s)

ELISA: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 5-15 μg/mL using human tissue
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANGPT2(285)

General description

Angiopoietin 2 is an endothelial cell-specific protein and has a pivotal role in mature blood vessel formation, endothelial cell proliferation and embryogenic angiogenesis. Angiopoietin 2 produced by endothelial cells of interior tumor vessels, stimulates vessel regression and disassociation from surrounding tumor cells. Anti-angiopoietin-2 antibody can be used in western blotting. Goat anti-angiopoietin-2 antibody reacts specifically with recombinant human angiopoietin-2. The product may also show 25% (approx.) cross-reactivity for recombinant human angiopoietin-1.

Immunogen

purified recombinant human angiopoietin-2 expressed in mouse NSO cells.

Application

Anti-angiopoietin-2 antibody can be used in immunohistochemistry for detecting humanangiopoietin-2 using paraffin-embedded human tissue sections. It can also be used in ELISA.
Goat anti-angiopoietin-2 antibody is also suitable for use in western blot assays at 0.1-0.2 μg/mL

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline (pH 7.4) with 5% trehalose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shigetomo Fukuhara et al.
Histology and histopathology, 25(3), 387-396 (2010-01-08)
Angiopoietin (Ang) 1 is a ligand for endothelium-specific receptor tyrosine kinase Tie-2. In adult vasculature, Ang1/Tie2 signaling is thought to regulate both maintenance of vascular quiescence and promotion of angiogenesis. However, it has been unknown how Tie2 signal regulates these
Jawun Choi et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 117, 109091-109091 (2019-06-23)
Diosmetin is a natural flavonoid obtained from citrus fruits and some medicinal herbs. Previous studies have reported the anti-cancer activity of diosmetin in some types of tumors. However, it is still unclear whether diosmetin exerts anti-cancer effects, particularly anti-angiogenic effects
Z Zhou et al.
Cell death and differentiation, 22(3), 488-498 (2015-01-13)
Damage and loss of the postmitotic photoreceptors is a leading cause of blindness in many diseases of the eye. Although the mechanisms of photoreceptor death have been extensively studied, few studies have addressed mechanisms that help sustain these non-replicating neurons

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service