Skip to Content
Merck
All Photos(1)

Key Documents

938B

Sigma-Aldrich

Diamond: Antibody Diluent

TBS, <1% BSA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

Quality Level

description

[For In Vitro Diagnostic Use in Select Regions]

form

buffered aqueous solution (blue)

packaging

pkg of 1 L ready to use (938B-09)
pkg of 200 mL ready to use (938B-05)
pkg of 50 mL ready to use (938B-03)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Diamond: Antibody Diluent is an appropriately buffered solution for the dilution of both polyclonal and monoclonal antibodies as well as for the preparation of negative control reagents in IHC. It is designed to minimize the non-specific reaction that may be caused by the antibody and encourage specific antigen-antibody binding. Diamond: Antibody Diluent can also be used to stabilize diluted antibodies when stored at 2-8°C.

Physical form

Solution containing Tris buffer, pH 7.3-7.7, with 1% BSA and <0.1% sodium azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service