Skip to Content
Merck
All Photos(1)

Key Documents

93362

Sigma-Aldrich

Trizma® base

≥99.8% (T), crystalline, for molecular biology, BioUltra

Synonym(s):

2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
Beilstein:
741883
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.25

Product Name

Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)

grade

Molecular Biology
for molecular biology

description

aminopeptidase substrate

product line

BioUltra

Assay

≥99.8% (T)

form

crystalline

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

ign. residue

≤0.01% (as SO4)

loss

≤0.5% loss on drying, 110 °C

color

colorless to white

pH

10.5-12.0 (25 °C, 4 M in H2O)

useful pH range

7-9

pKa (25 °C)

8.1

bp

219-220 °C/10 mmHg (lit.)

mp

167-172 °C (lit.)
168-172 °C

solubility

H2O: 4 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤20 mg/kg
sulfate (SO42-): ≤5 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

NC(CO)(CO)CO

λ

4 M in H2O

UV absorption

λ: 260 nm Amax: 0.10
λ: 280 nm Amax: 0.08

suitability

in accordance for luminescence

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris is an established basimetric standard and buffer used in biochemistry and molecular biology. It may be used by itself as a buffer or as a component of mixed buffer formulations, such as Tris-EDTA (TE) buffer, Tris-acetate-EDTA (TAE) buffer, Tris-borate-EDTA (TBE) buffer, etc. It is pure, essentially stable, relatively non-hygroscopic and has a high equivalent weight.

The BioChemika quality is for the usual biochemical applications. The products designated as BioChemika Ultra grade are suitable for different applications like purification, precipitation, crystallization and other applications which require tight control of elemental content. Trace elemental analyses have been performed for all qualities. The molecular biology qualities are also tested for absence of nucleases and the luminescence quality spectroscopic tests are performed.

Application

Trizma has been used:
  • In the dissection medium and culture medium of Hippocampal organotypic tissue cultures
  • As a component of lysis buffer before immunoprecipitation and immunoblotting
  • As a component of harvest buffer, lysis buffer, ChIP dilution buffer, and wash buffer in chromatin immunoprecipitation method for studying protein-DNA binding
  • As a component of mixed buffer formulations
  • As a component of NuPAGE loading buffer for SDS-PAGE and as a component of transfer buffer for Western blotting
Trizma is used in the formulation of buffer solutions in the pH range between 7.5 and 8.5. Tris buffer solutions are widely used in cell and molecular biology for processes such as protein and nucleic acid extraction and purification. Trizma based buffers are also in column chromatography and in gel electrophoresis. Trizma base is used as a general reagent for the preparation of all types of Tris buffers.

Other Notes

Easily compare specifications for Trizma products with the Trizma specification table.
Running buffer component in non-denaturing agarose gel electrophoresiscitation; In non-denaturing and urea-denaturing polyacrylamide gel electrophoresiscitation; DNA digest analysis with capillary electrophoresiscitation.
The pH values of all buffers are temperature- and concentration-dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution.
For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.
Running buffer component in non-denaturing agarose gel electrophoresis; In non-denaturing and urea-denaturing polyacrylamide gel electrophoresis; DNA digest analysis with capillary electrophoresis.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new method allowing long-term potentiation recordings in hippocampal organotypic slices.
Paci P, et al.
Brain and Behavior, 7(5), e00692-e00692 (2017)
A fast, efficient chromatin immunoprecipitation method for studying protein-DNA binding in Arabidopsis mesophyll protoplasts.
Plant methods, 13(1), 42-42 (2017)
Sambrook J and Russell DW
Molecular Cloning: A Laboratory Manual (2001)
Differentiation of ruminant transmissible spongiform encephalopathy isolate types, including bovine spongiform encephalopathy and CH1641 scrapie.
Jacobs JG, et al.
The Journal of General Virology, 92(1), 222-232 (2011)
In vitro Detection of S-acylation on Recombinant Proteins via the Biotin-Switch Technique.
Qi D and Innes RW
Bio-protocol, 4(22) (2014)

Protocols

Annealing is the process of heating and cooling two single-stranded oligonucleotides with complementary sequences.

Annealing is the process of heating and cooling two single-stranded oligonucleotides with complementary sequences.

Annealing is the process of heating and cooling two single-stranded oligonucleotides with complementary sequences.

Annealing is the process of heating and cooling two single-stranded oligonucleotides with complementary sequences.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service