90469
(RS)-Mevalonic acid lithium salt
≥96.0% (GC)
Synonym(s):
(±)-MVA-Li, rac.-MVA-Li, Lithium (±)-3,5-dihydroxy-3-methyl-pentanoate, Lithium (±)-3,5-dihydroxy-3-methyl-valerate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H11O4 · xLi+
CAS Number:
Molecular Weight:
147.15 (free acid basis)
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.25
Assay:
≥96.0% (GC)
form:
solid
Recommended Products
Quality Level
Assay
≥96.0% (GC)
form
solid
composition
lithium, 2.2-4.7%
storage temp.
2-8°C
SMILES string
OC(CCO)(CC(=O)O)C
InChI
1S/C6H12O4/c1-6(10,2-3-7)4-5(8)9/h7,10H,2-4H2,1H3,(H,8,9)
InChI key
KJTLQQUUPVSXIM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Metabolite of the mevalonate pathway, which plays a key role in the biosynthesis of sterols, dolichol, heme and ubiquinone. Of interest for research in the disease areas oncology, autoimmune diseases, artherosclerosis and Alzheimer disease, as well as for inherited deficiencies of mevalonate kinase.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Martin Thurnher et al.
Biochimica et biophysica acta, 1831(6), 1009-1015 (2013-03-26)
The mevalonate pathway is a highly conserved metabolic cascade and provides isoprenoid building blocks for the biosynthesis of vital cellular products such as cholesterol or prenyl pyrophosphates that serve as substrates for the posttranslational prenylation of numerous proteins. The pathway
Ai Hosaka et al.
Neurochemical research, 38(3), 589-600 (2012-12-28)
Statins, 3-hydroxy-3-methylglutaryl-coenzyme A (HMG-CoA) reductase inhibitors, have been reported to attenuate amyloid-β peptide (Aβ) production in various cellular models. However, the mechanisms by which statins affect neuronal Aβ production have not yet been clarified. Here, we investigated this issue in
Adelene Ai-Lian Song et al.
PloS one, 7(12), e52444-e52444 (2013-01-10)
Isoprenoids are a large and diverse group of metabolites with interesting properties such as flavour, fragrance and therapeutic properties. They are produced via two pathways, the mevalonate pathway or the 2-C-methyl-D-erythritol-4-phosphate (MEP) pathway. While plants are the richest source of
Jordi Pérez-Gil et al.
The Biochemical journal, 452(1), 19-25 (2013-04-26)
Isoprenoids are a large family of compounds synthesized by all free-living organisms. In most bacteria, the common precursors of all isoprenoids are produced by the MEP (methylerythritol 4-phosphate) pathway. The MEP pathway is absent from archaea, fungi and animals (including
Jianxin Shi et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 34(1), 429-435 (2012-11-28)
Esophageal squamous cell carcinoma (ESCC) is one of the most common lethal tumors in the world. Thus, it is very urgent to develop new therapeutic targets against this disease. The mevalonate (MVA) pathway, paced by its rate-limiting enzyme, hydroxymethylglutaryl coenzyme
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service