Skip to Content
Merck
All Photos(1)

Key Documents

86614

Sigma-Aldrich

Tetraethylammonium chloride

BioUltra, for molecular biology, ≥99.0% (AT)

Synonym(s):

Etamon chloride, TEAC, TEA chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(Cl)
CAS Number:
Molecular Weight:
165.70
Beilstein:
3563247
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.26

grade

for molecular biology

Quality Level

product line

BioUltra

Assay

≥99.0% (AT)

form

powder

mol wt

165.70 g/mol

technique(s)

gas chromatography (GC): suitable

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

bromide (Br-): ≤500 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.03
λ: 280 nm Amax: 0.03

SMILES string

[Cl-].CC[N+](CC)(CC)CC

InChI

1S/C8H20N.ClH/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H/q+1;/p-1

InChI key

YMBCJWGVCUEGHA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetraethylammonium (TEA), a simple quaternary ammonium compound, is commonly used to block potassium channels. It can physically enter the pore and block the channel. Additionally, TEA either imitates or antagonizes the actions of choline and acetylcholine, two other quaternary ammonium ions.

Application

Tetraethylammonium chloride has been used:

  • as a pharmacological agent in electrophysiology to study its pharmacological effects on Cnidarians
  • as a component in refolding buffer for protein refolding
  • to puff onto isolated patches to block voltage-gated K+ (Kv3) channels and post-hoc evaluation

Features and Benefits

  • Assay: ≥99.0% (AT), BioUltra, for molecular biology
  • Suitable for gas chromatography (GC)

Other Notes

TEACl method of DNA-DNA hybridization

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A. Caccone et al.
NATO ASI Ser., Ser H, 57, 385-385 (1991)
Viktor J Olah et al.
eLife, 11 (2022-06-22)
In Alzheimer's disease (AD), a multitude of genetic risk factors and early biomarkers are known. Nevertheless, the causal factors responsible for initiating cognitive decline in AD remain controversial. Toxic plaques and tangles correlate with progressive neuropathology, yet disruptions in circuit
Avinash Jaiganesh et al.
Structure (London, England : 1993), 26(9), 1210-1225 (2018-07-24)
Cadherin-23 (CDH23) is an essential component of hair-cell tip links, fine filaments that mediate inner-ear mechanotransduction. The extracellular domain of CDH23 forms about three-fourths of the tip link with 27 extracellular cadherin (EC) repeats that are structurally similar but not
Tetraethylammonium
Simmons MA
xPharm: The Comprehensive Pharmacology Reference, 1-3 (2007)
Timothy S Balmer et al.
eLife, 10 (2021-02-23)
Synapses of glutamatergic mossy fibers (MFs) onto cerebellar unipolar brush cells (UBCs) generate slow excitatory (ON) or inhibitory (OFF) postsynaptic responses dependent on the complement of glutamate receptors expressed on the UBC's large dendritic brush. Using mouse brain slice recording

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service