Skip to Content
Merck
All Photos(1)

Key Documents

85L2

Sigma-Aldrich

Leukocyte Alkaline Phosphatase Kit

based on naphthol AS-MX phosphate and fast blue RR salt (with citrate)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.47

Quality Level

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

dilution

(for histology)

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Leukocyte Alkaline Phosphatase (LAP) kits are intended for the qualitative demonstration of alkaline phosphatase activity in white blood cells.
Peripheral blood or bone marrow preparations are fixed to a microscope slide. The film is then incubated in a mixture of naphthol AS-MX phosphate alkaline solution with fast blue RR salt. The resulting blue, insoluble, granular dye deposit indicates sites of alkaline phosphatase activity.

Kit Components Also Available Separately

Product No.
Description
SDS

  • MHS1Mayer′s Hematoxylin Solution 100 mLSDS

  • 855Naphthol AS-MX Phosphate Alkaline Solution, 0.25% 20 mLSDS

  • 854CCitrate Concentrated Solution 20 mLSDS

  • FBS25Fast Blue RR Salt, Preweighed Capsules 10 capsulesSDS

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Bouet et al.
European cells & materials, 29, 250-266 (2015-05-16)
An engineered three dimensional (3D) in vitro cell culture system was designed with the goal of inducing and controlling in vitro osteogenesis in a reproducible manner under conditions more similar to the in vivo bone microenvironment than traditional two-dimensional (2D)
Clarissa R Taufer et al.
The International journal of developmental biology, 64(7-8-9), 433-443 (2020-10-17)
The neural crest (NC) is a transitory embryonic structure of vertebrates that gives rise to an astonishing variety of derivatives, encompassing both neural and mesenchymal cell types. Neural crest cells (NCCs) are an excellent model to study how environmental factors
Luca Azzolin et al.
Cell, 151(7), 1443-1456 (2012-12-19)
Wnt growth factors are fundamental regulators of cell fate, but how the Wnt signal is translated into biological responses is incompletely understood. Here, we report that TAZ, a biologically potent transcriptional coactivator, serves as a downstream element of the Wnt/β-catenin
Guenaelle Bouet et al.
PloS one, 10(2), e0117402-e0117402 (2015-02-25)
Bone sialoprotein (BSP) belongs to the "small integrin-binding ligand N-linked glycoprotein" (SIBLING) family, whose members interact with bone cells and bone mineral. BSP is strongly expressed in bone and we previously showed that BSP knockout (BSP-/-) mice have a higher
Haibo Zhao et al.
Developmental cell, 14(6), 914-925 (2008-06-10)
Maintenance of bone mass and integrity requires a tight balance between resorption by osteoclasts and formation by osteoblasts. Exocytosis of functional proteins is a prerequisite for the activity of both cells. In the present study, we show that synaptotagmin VII

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service