Skip to Content
Merck
All Photos(1)

Key Documents

85976

Sigma-Aldrich

Methoxypolyethylene glycol succinate N-hydroxysuccinimide

5,000, ≥90%

Synonym(s):

Polyethylene glycol, mono-Methyl polyethylene glycol succinate N-succinimidyl ester, O-Methyl-O′-succinylpolyethylene glycol 5,000 N-succinimidyl ester, PEG-succinimidyl succinate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥90%

form

powder

storage temp.

−20°C

SMILES string

N1(C(=O)CCC1=O)OC(=O)CCC(=O)COCCO

InChI

1S/C11H15NO7/c13-5-6-18-7-8(14)1-4-11(17)19-12-9(15)2-3-10(12)16/h13H,1-7H2

InChI key

OVPTZXRXNNJKSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Matrix metalloproteinases-2/9-sensitive peptide-conjugated polymer micelles for site-specific release of drugs and enhancing tumor accumulation: preparation and in vitro and in vivo evaluation. This study explores the use of Methoxypolyethylene glycol succinate N-hydroxysuccinimide (MPEG-S-NHS) for creating peptide-conjugated polymer micelles that are sensitive to matrix metalloproteinases-2/9. These micelles are designed for site-specific drug release and increased tumor targeting, showcasing a promising application in advanced drug delivery systems (Zhang X, Wang X, Zhong W, Ren X, Sha X, Fang X, 2016) [link].


Linkage

Shearwater Polymers product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Croyle et al.
Journal of virology, 75(10), 4792-4801 (2001-04-20)
Most of the early gene therapy trials for cystic fibrosis have been with adenovirus vectors. First-generation viruses with E1a and E1b deleted are limited by transient expression of the transgene and substantial inflammatory responses. Gene transfer is also significantly curtailed
Y P Li et al.
Acta pharmacologica Sinica, 22(6), 549-555 (2001-12-19)
To assess the merits of polyethylene glycol-modified recombinant human tumor necrosis factor alpha (PEG-rHuTNF-alpha). The rHuTNF-alpha was modified with N-succinimidyl succinate monomethoxy polyethylene glycol (SS-PEG) of three different molecular weights. The PEG-rHuTNF-alpha was separated into fractions of various molecular weights
G Sirokmán et al.
Protein science : a publication of the Protein Society, 2(7), 1161-1170 (1993-07-01)
Bacteriorhodopsin (BR), from the purple membrane (PM) of Halobacterium halobium, was chemically modified with methoxypolyethylene glycol (m-PEG; molecular weight = 5,000 Da) succinimidyl carbonate. The polyethylene glycol-bacteriorhodopsin (m-PEG-SC-BR33) conjugate, containing one polyethylene glycol chain, was water soluble. The secondary structure
H Matsuyama et al.
Chemical & pharmaceutical bulletin, 39(3), 743-746 (1991-03-01)
Phospholipase D from Streptomyces sp. AA586, PLDP, was modified with methoxypolyethylene glycol succinimidylsuccinate (ss-PEG), an active derivative of polyethylene glycol. By titration with trinitrobenzene sulfonate (TNBS), approximately 70% of the free amino groups in the enzyme protein were shown to
Georgi As Georgiev et al.
Colloids and surfaces. B, Biointerfaces, 59(2), 184-193 (2007-06-26)
Foam thin liquid films (TLF) and monolayers at the air-water interface formed by DMPC mixed with DMPE-bonded poly (ethylene glycol)s (DMPE-PEG(550), DMPE-PEG(2000) and DMPE-PEG(5000)) were obtained. The influence of both (i) PEG chain size (evaluated in terms of Mw) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service