Skip to Content
Merck
All Photos(1)

Documents

82435

Sigma-Aldrich

Puerarin

≥98.0% (HPLC)

Synonym(s):

8-(β-D-Glucopyranosyl-7-hydroxy-3- (4-hydroxyphenyl)-4H-1-benzopyran-4-one, Kakonein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O9
CAS Number:
Molecular Weight:
416.38
Beilstein:
64198
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98.0% (HPLC)

form

powder

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)ccc3C(=O)C(=COc23)c4ccc(O)cc4

InChI

1S/C21H20O9/c22-7-14-17(26)18(27)19(28)21(30-14)15-13(24)6-5-11-16(25)12(8-29-20(11)15)9-1-3-10(23)4-2-9/h1-6,8,14,17-19,21-24,26-28H,7H2/t14-,17-,18+,19-,21+/m1/s1

InChI key

HKEAFJYKMMKDOR-VPRICQMDSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Isoflavone component of kudzu. Reduces anxiety symptoms associated with alcohol withdrawal.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

David H Overstreet et al.
Pharmacology, biochemistry, and behavior, 75(3), 619-625 (2003-08-05)
Because extracts of kudzu have been used as a hangover remedy in China for many centuries, we tested the ability of NPI-031G (puerarin), an isoflavone isolated from kudzu, to counteract anxiogenic effects associated with withdrawal from chronic alcohol exposure. NPI-O31G
Xiang-Yang Sun et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 35(6), 980-983 (2012-12-15)
To optimize the parameters of extraction technology for Puerarin from Pueraria lobata by Central Composite Design-Response Surface Methodology. Used the ratio of dosage liquid and extraction time as the examination factor,the content of puerarin for the extraction percentage as the
Rong Li et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 52, 69-75 (2012-11-14)
This study was designed to evaluate the potential effects of puerarin (PR), an effective isoflavonoid compound purified from Pueraria lobata, in treating hepatic fibrosis (HF) rats induced by carbon tetrachloride (CCl(4), 2 mL kg(-1) d(-1)). Compared to model control, PR
Y Zou et al.
Free radical research, 47(1), 55-63 (2012-10-24)
Current evidence suggests that amyloid beta (Aβ) peptides may play a major role in the pathogenesis of Alzheimer's disease in part by eliciting oxidative stress. Puerarin, a major isoflavone glycoside from Kudzu root (Pueraria lobata), has been reported to exert
Lin Wang et al.
Human & experimental toxicology, 32(2), 172-185 (2013-01-15)
Puerarin (PU), a natural flavonoid, has been reported to possess anti-oxidative and anti-inflammatory activities. In the present study, female Sprague-Dawley rats received lead (Pb) nitrate (300 mg/L, via drinking water) and/or PU (400 mg/kg/day, orally) to investigate the protective effects of PU

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service