Skip to Content
Merck
All Photos(1)

Key Documents

79480

Sigma-Aldrich

D-2-Phosphoglyceric acid barium salt hydrate

≥70% (calc. on dry substance, enzymatic), powder

Synonym(s):

2-PG-Ba, D-Glycerate 2-phosphate barium salt, Barium D-2-phosphoglycerate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5BaO7P · xH2O
CAS Number:
Molecular Weight:
321.37 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.26

Product Name

D-2-Phosphoglyceric acid barium salt hydrate, ≥70% (calc. on dry substance, enzymatic)

Assay

≥70% (calc. on dry substance, enzymatic)

form

powder

impurities

≤35% water

storage temp.

2-8°C

SMILES string

[Ba+2].[P](=O)([O-])([O-])O[C@H](CO)C(=O)O

InChI

1S/C3H7O7P.Ba/c4-1-2(3(5)6)10-11(7,8)9;/h2,4H,1H2,(H,5,6)(H2,7,8,9);/q;+2/p-2/t2-;/m1./s1

InChI key

VDCXVAFBCOMLAH-HSHFZTNMSA-L

General description

D-2-Phosphoglyceric acid barium salt hydrate is a substrate for the assay of 3-phosphoglyceric acid mutase; and enolase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Oesper
Methods in Enzymology, 1, 423-423 (1955)
Dynamic fluctuations in a bacterial metabolic network.
Bi, et al.
Nature Communications, 14, 2173-2173 (2023)
T. Bucher
Methods in Enzymology, 427-427 null
Zeng Li et al.
Vaccines, 8(3) (2020-07-30)
Trypanosoma evansi is a widely spread parasite that causes the debilitating disease "surra" in several types of ungulates. This severely challenges livestock rearing and heavily weighs on the socio-economic development in the affected areas, which include countries on five continents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service