Skip to Content
Merck
All Photos(1)

Key Documents

76085

Sigma-Aldrich

Anti-Mouse IgG - Atto 594 antibody produced in goat

~1 mg/mL protein, affinity isolated antibody

Synonym(s):

Atto 594 - goat-Anti-mouse IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

Atto 594 conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

contains

50% glycerol as stabilizer

species reactivity

mouse

concentration

~1 mg/mL protein

fluorescence

λex 603 nm; λem 625 nm in PBS

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

General description

Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases. Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice.
Affinity isolated antigen specific antibody is purified from goat anti-mouse IgG antiserum to remove essentially all goat serum proteins, including immunoglobulin. Goat anti-mouse IgG associates with mouse IgGs.

Immunogen

purified mouse IgG

Application

Atto 594-goat anti-mouse IgG can be used for fluorescence-based immunoassays.

Analysis Note

unconjugated dye ≤5% of total fluorescence; dye-to-protein ratio ≥2

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles Bou-Nader et al.
Cell host & microbe, 29(9), 1421-1436 (2021-08-14)
The HIV-1 virion structural polyprotein, Gag, is directed to particle assembly sites at the plasma membrane by its N-terminal matrix (MA) domain. MA also binds to host tRNAs. To understand the molecular basis of MA-tRNA interaction and its potential function
Krzysztof Marycz et al.
International journal of nanomedicine, 16, 6049-6065 (2021-09-14)
Healing of osteoporotic defects is challenging and requires innovative approaches to elicit molecular mechanisms promoting osteoblasts-osteoclasts coupling and bone homeostasis. Cytocompatibility and biocompatibility of previously characterised nanocomposites, i.e Ca5(PO4)3OH/Fe3O4 (later called nHAp/IO) functionalised with microRNAs (nHAp/IO@miR-21/124) was tested. In vitro
Paola Muttathukunnel et al.
Proceedings of the National Academy of Sciences of the United States of America, 119(45), e2119044119-e2119044119 (2022-11-03)
Robust neural information transfer relies on a delicate molecular nano-architecture of chemical synapses. Neurotransmitter release is controlled by a specific arrangement of proteins within presynaptic active zones. How the specific presynaptic molecular architecture relates to postsynaptic organization and how synaptic
Mirko Cortese et al.
Cell host & microbe, 28(6), 853-866 (2020-11-28)
Pathogenesis induced by SARS-CoV-2 is thought to result from both an inflammation-dominated cytokine response and virus-induced cell perturbation causing cell death. Here, we employ an integrative imaging analysis to determine morphological organelle alterations induced in SARS-CoV-2-infected human lung epithelial cells.
Anu G Nair et al.
Cell reports, 37(11), 110105-110105 (2021-12-16)
Presynaptic homeostatic plasticity (PHP) stabilizes synaptic transmission by counteracting impaired neurotransmitter receptor function through neurotransmitter release potentiation. PHP is thought to be triggered by impaired receptor function and to involve a stereotypic signaling pathway. However, here we demonstrate that different

Articles

Immunoblotting (Western blot transfer) is a common technique in modern proteomics research.

Protocols

TrueGel3D® Hydrogel Plate protocol guides high-throughput culture of human adipose MSCs for screening applications.

TrueGel3D® Hydrogel Plate protocol guides high-throughput culture of human adipose MSCs for screening applications.

TrueGel3D® Hydrogel Plate protocol guides high-throughput culture of human adipose MSCs for screening applications.

TrueGel3D® Hydrogel Plate protocol guides high-throughput culture of human adipose MSCs for screening applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service