Skip to Content
Merck
All Photos(1)

Key Documents

72091

Sigma-Aldrich

NBT-BCIP® solution

BioReagent

Synonym(s):

BCIP®/NBT, NBT-X-phosphate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

product line

BioReagent

Quality Level

form

liquid

technique(s)

microbe id | staining: suitable

solubility

0.1 M Tris-HCl, pH 7.4: 0.0175 mL/mL, clear

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].Cc1ccc(N)cc1.OP(O)(=O)Oc2c[nH]c3ccc(Br)c(Cl)c23.COc4cc(ccc4-[n+]5nc(nn5-c6ccc(cc6)[N+]([O-])=O)-c7ccccc7)-c8ccc(c(OC)c8)-[n+]9nc(nn9-c%10ccc(cc%10)[N+]([O-])=O)-c%11ccccc%11

InChI

1S/C40H30N10O6.C8H6BrClNO4P.C7H9N.2ClH/c1-55-37-25-29(13-23-35(37)47-43-39(27-9-5-3-6-10-27)41-45(47)31-15-19-33(20-16-31)49(51)52)30-14-24-36(38(26-30)56-2)48-44-40(28-11-7-4-8-12-28)42-46(48)32-17-21-34(22-18-32)50(53)54;9-4-1-2-5-7(8(4)10)6(3-11-5)15-16(12,13)14;1-6-2-4-7(8)5-3-6;;/h3-26H,1-2H3;1-3,11H,(H2,12,13,14);2-5H,8H2,1H3;2*1H/q+2;;;;/p-2

InChI key

GDPIIVBVMSORRQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Physical form

solution of 18.8 mg/ml nitro-blue tetrazolium chloride; 9.4 mg/ml 5-bromo-4-chloro-3-indolylphosphate toluidine salt in 67% DMSO

Other Notes

Chromogenic phosphatase substrate producing a blue-colored precipitate at the site of enzymatic activity[1]

Legal Information

BCIP is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Makrina Totsika et al.
Journal of bacteriology, 191(12), 3901-3908 (2009-04-21)
Disulfide bond (DSB) formation is catalyzed by disulfide bond proteins and is critical for the proper folding and functioning of secreted and membrane-associated bacterial proteins. Uropathogenic Escherichia coli (UPEC) strains possess two paralogous disulfide bond systems: the well-characterized DsbAB system
Andrea L H Arnett et al.
Frontiers in microbiology, 2, 220-220 (2011-11-09)
Recombinant adeno-associated viral (rAAV) vectors promote long-term gene transfer in many animal species. Significant effort has focused on the evaluation of rAAV delivery and the immune response in both murine and canine models of neuromuscular disease. However, canines provided for
Ludovic Sauguet et al.
Nucleic acids research, 39(10), 4475-4489 (2011-02-08)
Cyclodipeptide synthases (CDPSs) belong to a newly defined family of enzymes that use aminoacyl-tRNAs (aa-tRNAs) as substrates to synthesize the two peptide bonds of various cyclodipeptides, which are the precursors of many natural products with noteworthy biological activities. Here, we
Dariusz Abramczyk et al.
Preparative biochemistry & biotechnology, 39(3), 277-288 (2009-05-12)
The class V chitin synthase is unique because it has a myosin motor-like domain fused to its catalytic domain. The biochemical properties of this enzyme and its function remain undefined beyond the knowledge that it is the only single chitin
Ying Wu et al.
Infection and immunity, 76(5), 2249-2255 (2008-03-12)
The 10-kDa culture filtrate protein (CFP-10) and 6-kDa early secretory antigen of T cells (ESAT-6) are secreted in abundance by Mycobacterium tuberculosis and are frequently recognized by T cells from infected people. The genes encoding these proteins have been deleted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service