Skip to Content
Merck
All Photos(1)

Key Documents

61668

Sigma-Aldrich

rac-Glycerol 1-phosphate sodium salt hydrate

≥95.0% (TLC)

Synonym(s):

α-Glycerol phosphate sodium salt hydrate, DL-Glycerol-1-phosphate sodium salt hydrate, DL-Glycerol-3-phosphate sodium salt hydrate, Sodium 3-phosphoglycerate hydrate, Sodium DL-glycerophosphate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H9O6P · xNa+ · yH2O
CAS Number:
Molecular Weight:
172.07 (anhydrous free acid basis)
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥95.0% (TLC)
95-102% anhydrous basis (corrected for sodium, T)

form

powder or solid

impurities

≤35% water

storage temp.

2-8°C

SMILES string

OCC(O)COP(O)(O)=O

InChI

1S/C3H9O6P/c4-1-3(5)2-9-10(6,7)8/h3-5H,1-2H2,(H2,6,7,8)

InChI key

AWUCVROLDVIAJX-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical studies of the protective action of phosphate compounds against the demineralization of human dental enamel in vitro.
T H Grenby et al.
Caries research, 14(4), 210-220 (1980-01-01)
Response surface optimization of the medium components for the production of biosurfactants by probiotic bacteria.
Rodrigues,L., et al.
Process Biochemistry (Oxford, United Kingdom), 41, 1-10 (2006)
Utilization of dissolved organic phosphorus by different groups of phytoplankton taxa.
Wang, Z.H., et al.
Harmful Algae, 12, 113-118 (2011)
MALDI-TOF MS analysis of the extracellular polysaccharides released by the diatom Thalassiosira pseudonana under various nutrient conditions.
Ai, X.X.et al.
Journal of Applied Phycology, 27, 673-684 (2015)
A novel mono- and diacylglycerol lipase highly expressed in Pichia pastoris and its application for food emulsifier preparation.
Huang, J., et al.
Process Biochemistry (Oxford, United Kingdom), 48, 1899-1904 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service